Check list for scale balance
Belt conveyer scales have become an important part of most bulk material handling facilities, including at mines all over the world. However, being relatively unobtrusive in most conveyor systems, they are often overlooked or ignored after their initial installation.
Does your big tough equipment really need tender loving care? Most definitely.
Lack of simple maintenance will potentially cause a significant reduction in the accuracy capabilities of these instruments. Most belt conveyor scales are capable of providing reliable results within ±½% of their full-scale rating. Belt conveyor scales that have been certified via the National Type Evaluation Program (NTEP) are capable of providing results that meet the criteria of the National Institute of Standards and Technology (NIST) Handbook-44, which is ¼%. This is defined as being within ¼% of actual material load and repeatable within a ¼% bandwidth.
Regardless of the accuracy capability of the scale design, it is unlikely that these devices will perform as promised if simple maintenance procedures are not strictly adhered to. Every scale installation will develop its own set of operating characteristics, so it is absolutely necessary to monitor the scale’s performance and provide routine maintenance as required.
Throughout the belt scale industry, it is advised that calibration checks be made frequently during the weeks after initial installation, then to increase the time frame between calibrations as statistical results are obtained. While this is a simple suggestion to follow, all too frequently the calibration verification time frames expand until the point where they are considered only when a problem is suspected. But by that time it is too late. What’s the big deal? By the time a problem is noticed, incorrect weighments have been made, processes have been interrupted and inventory levels had to be verified by other means.
These problems can be avoided with the simple establishment of a routine inspection procedure, including not only the belt conveyor scale, but the entire material handling system. Any changes that occur or are performed within the conveyor can and probably will affect the performance of the scale, which of course can affect the whole system.
Below is a quick recommended Maintenance Check List chart. Use it as a guide to help ensure your belt conveyer scales are not throwing your bulk material handling facilities off-balance.
Make sure you read the next article: “Idler Alignment on Belt Conveyor Scales”
VERIFICATION INTERVAL | ||||||
ITEM | DAILY | WEEKLY | MONTHLY | QUARTERLY | ANNUALLY | NOTES |
scale area – debrisX | X | Clean scale area. Determine cause of debris and take steps to remedy. | ||||
zero cal | X | Perform Auto Zero procedure. Accept and record any changes. If change is > .25%, identify cause and correct. Record results | ||||
idler rolls condition | X | Inspect idlers for wear / damage. Replace rolls or bearings as needed. | ||||
span cal | X | Perform Auto SPAN simulated load tests. Check repeatability. Record results. Refer the HB-44 section UR.3.2 for actions if error exceeds .25%. | ||||
belt scraper | X | Check operation, adjust or replace blades if worn. | ||||
belt condition | X | Visual inspection for cuts, tears or worn edges. | ||||
take-up | X | Inspect for free travel (bearings, sheaves, etc.) | ||||
speed pulley | X | Inspect for wear, material build-up, belt wrap. Check bearings. | ||||
speed sensor coupling | X | Inspect for tightness, wobble and corrosion. | ||||
load cell offset | X | No load output must be within 1% of rated maximum (typical “S” cell is +/- .3 mV) | ||||
load cell balance | X | Multiple load cell scales must be balanced to within 1 mV | ||||
static weight condition | X | Check for corrosion, location and clearances | ||||
revolution time | X | Verify time for 1 belt rev at maximum speed. | ||||
zero reference number | X | Compare ZERO # with Reference #. Maximum change is 2% / year | ||||
audit trail | X | Review scale history | ||||
line voltage | X | Measure Hot to Neutral, Hot to Ground, Neutral to Ground. Correct as needed. | ||||
alignment | X | Complete per instruction manual | ||||
excitation | X | Verify value and stability | ||||
belt length | X | Measure and verify. Perform acquire test duration if changes noted. | ||||
check rods | X | Inspect check rods. Rods must be straight, spherical washers w/o corrosion. | ||||
I/O integrity | X | Check and verify performance of all I/O being used. | ||||
dead band | X | Confirm settings and adjust if necessary | ||||
auto zero track limit | X | Record Data | ||||
auto zero track correction | X | Record Data | ||||
passwords | X | Confirm and revise if required | ||||
wire terminations | X | Inspect for corrosion and tightness | ||||
cable integrity | X | Visual and ohm check. (corrosion, moisture, deterioration) | ||||
spherical washers | X | Inspect for corrosion, pitting, etc. Replace if necessary. | ||||
material factors | X | Verify with weighed load test. | ||||
*After performing ANY tests or maintenance as part of weekly, monthly, quarterly or annual schedule, ALWAYS repeat zero and span tests ! | ||||||
Leave a Reply