New to the Mining Industry? Make Sure You Know the Most Common Types of Mining Equipment
mining equipmentWorking in the mining industry can be a dangerous place if you don’t know what you’re doing. Regular training is essential and understanding the machines you’re working near or operating plays an important role in all aspects of the industry.
Each type of mining equipment comes with its own set of mining activities. The most common types of mining equipment vary depending whether the work is being carried out above or below ground or mining for gold, metals, coal or crude oil. From drilling machines to excavators, crushing and grinding equipment – the mining industry comes complete with all the right tools. New to the job and want to find out what it all means? Here’s a few of the industry’s most common types of equipment and why they’re important for the job.
Mining Drills
Probably one of the most common pieces of mining equipment, drills are an important part of the underground mining operation. Underground mining is carried out when rocks or minerals are located at a fair distance beneath the ground. But then they need to be brought to the surface. Underground specialized mining equipment such as trucks, loaders, diggers etc. are used to excavate the material and are normally hauled to the surface with skips or lifts for further processing. Drilling is normally required to place explosive charges to liberate the minerals from the overburden material. Underground mining techniques have progressed significantly over the past years, including using remote controlled machinery.
Drills assist in creating holes descending underground. If miners are required to work underground, drills can also be used in ensuring the holes are large enough to serve as a portal for miners to enter. Directional drilling is also a type of mining technology where miners will use the tools and certain methods to drill wells.
Blasting Tools 
Blasting tools are an essential part of the mining industry and are used to break down and fracture materials (usually rocks) by use of a calculated amount of explosive to liberate the sought-after product from the waste material. Blasting is also used to remove pockets of unwanted material that are preventing mining machines and personnel to get to the seam containing the materials of interest. Unmanned drill rigs will drill holes at pre-determined depths and positions on a blast face to ensure that a particular size fraction is achieved and that little of the overburden is liberated with the blasting to reduce material handling costs.  Once this process has been completed, an excavator is used to recover the blasted rocks and other debris that has been dislodged during the blasting.  The material is then conveyed to a central conveying system which will take it directly to the surface or via a skip and hoist system.
Blasting equipment is used for both underground and open pit mining operations and is known to be one of the most hazardous aspects of the job.
For mining operations to be successful good blast designs are absolutely vital as poor practices and excessive explosives can result in damage to rock structures causing unwanted caving.
Earth Movers
For above ground mining, earth movers are utilised regularly to carry loose soil and earth from one location to another. Earth movers play an important role in the mining industry because the equipment is specifically designed to work on large earth-moving and mining projects for a faster and more practical process. Used for digging, pushing and transporting the earth, they require the specialised skills of an operator.
Earth movers are heavy mining equipment that the industry would struggle to survive without and work hand in hand with bulldozers. Earth movers are normally used for removing overburden or waste material, which enables the excavators to remove the material or mineral of interest. Bulldozers are used to move this overburden material around to create a working surface for other equipment such as haul trucks and excavators.
Crushing Equipment
As its name suggests, crushing equipment is used to crush rock and stone. Designed to achieve maximum productivity and high reduction rate, mining crushing equipment can come in a variety of different types for a range of jobs.
Crushing equipment is specially configured to break down the hard rock matter or gravel to a manageable size for transportation or conveying. They are valuable pieces of equipment in the industry because they reduce the costs associated with handling of larger sized material and also ensure efficient liberation of elements of interest in downstream processing of the material. In an opencast or strip mining operation, the run of mine (ROM) material is normally transported to the primary crusher by haul trucks, and in underground mining operations it is conveyed to the primary crusher. Crushing equipment is important to the mining process because it reduces the use of precious excavated resources and eliminates the amount of material on site.
Feeding, Conveying, and On-line Elemental Analysis Equipment
Once the excavator transporter brings the raw material to the crusher for processing, the feeding device feeds the material into the crusher and in return the material is screened and all oversized material is recirculated back to the crusher to ensure correct size fraction is obtained. This weighbelt ‘feeding’ equipment, usually referred to as “Weighfeeders,” conveys and controls the feedrate into the crusher to improve crusher efficiency.
Feeding and conveying equipment are necessary to the mining industry to move and control material flow within a mining and processing operation to facilitate efficient operation of equipment and determine operating rates and yields. In some instances secondary crushing is required prior to processing of the material. Once the material is at the correct size, fraction processing can occur which could include, milling, flotation, leaching etc.
Belt scale systems let you monitor production output and inventory, or regulate product loadout, while providing vital information for the effective management and efficient operation of your business.  There are elemental crossbelt analyzers that provide real-time quality analysis of critical process streams to facilitate sorting, blending and out-of-seam dilution control. While materials are on the troughed belt conveyor, an automatic sampling system (which could be single or multi-stage) can take a representative sample directly from the moving material stream.  (Take a look at this video to see how a sampling system works.)  Weighbelt Feeders that convey and control feedrate accurately and reliably can reduce material consumption, help maintain blend consistency, and increase profits.
Flow measurement systems provide continuous, real-time flow measurement of free-falling materials or dense phase, pneumatically conveyed bulk solids, which is important to ensure and maintain product quality and process efficiency.
All these common mining tools are crucial to the industry and assist in getting the job done efficiently and cost effectively.
Visit our Cement, Coal, and Minerals Learning Center for more information on tools used for mining exploration, processing, data management, lab analysis, and safety.
Author Bios
This article was co-written by Jayde Ferguson, who writes for Direct Mining – a leading supplier of premier mining equipment, products and services throughout Australia and the Asia Pacific region. Stephan Nel, Global Product/Applications Manager for Coal & Sinter at Thermo Fisher Scientific, co-wrote and edited the piece as well.
NOTE:
belt scale selection guideNeed a Belt scale system for your bulk material handling?  To help you decide which belt scale system is best for your mining operation, we’ve outlined the options in an easy-to-read belt scale system selection guide so you can decide which belt scale system is right for you.  Click on the image, take a look at the chart, and see if it helps you decide.
Comments
  1. Alex Trodder says
    You make a great point about how drills are a very common piece of mining equipment. Choosing the right kind of drill tip can make a big difference in efficiency. Many drill bits will be tipped with synthetic diamonds to ensure that they are as durable as possible regardless of the rock and substrate involved. Thanks for your post.
  2. Tom says
    It is true that working in the mining industry can be a dangerous place if we don’t know about the machinery that are useful in mining industry. I hope your post will help people to know about the most common types of mining equipment and how to use them properly. Thanks
  3. Bob Lowe says
    Thanks for the information. This really help to understand the different types of equipment you can expect to see in a mine. Drilling is such an important part to the mining process. Drilling and blasting go hand in hand to help break up the hard ground. After all you can gather the material your mining if you can’t get to it.
  4. Zequek Estrada says
    Understating what type of mining equipment like core boxes, seems like a good place to start for beginners. There’s really now way to do a job properly unless you’re familiar with the tools and equipment you’ll be using. It might also help with improving safety.
  5. CHIDI YOUNG says
    I AM ABOUT GOING IN TO MINING BUSINESS , ON MY RESEARCH ON HOW IT IS DONE BROUGHT ME TO THIS ARTICLE KUDOS TO Author Bios FOR THIS WONDERFUL HELP I GOT THROUGH UR POST AM GRATEFUL AND I NEED MORE GUIDE LINE TOWARD STONE MINING I AM CHIDI YOUNG
  6. Rachel Frampton says
    I like how you mentioned that drills are needed in the mining industry because these are going to be used in creating holes descending underground. It’s also intriguing to learn that the blasting tools are mainly used to break down and fracture the materials. Well, if I had a mining company, I would make sure to place industrial fans in there because these will provide air stream.
  7. Rarik Sam says
    Thank you for sharing useful information amazing wonderful awesome fantastic Your article impressed me very heartily. Please keep sharing.