VIDEO: Elemental Analysis for Cement
cement videoIn this animated video, you’ll see how continuous online analysis of raw materials used in making cement is done. The online elemental analyzer provides chemistry analysis every minute to allow quick and accurate changes to be made to the process, so consistent raw material quality results with minimal deviation. Well controlled kiln feed chemistry with low variability has lower energy requirements and creates stable operating conditions. Also included in the video is an explanation of Prompt Gamma Neutron Activation Analysis (PGNAA) and how gamma rays are used to provide an energy spectrum of the samples for accurate analysis. Watch the 4-minute Elemental Analyzer for Cement Video.
Visit the Cement Production center on our website, with free resources to help you optimize your cement making process.