High-Quality TEM Lamella Preparation: Critical Factors and Best Practices
Introduction: The quality of transmission electron microscopy (TEM) lamella preparation directly determines the accuracy and reliability of semiconductor characterization, metrology, and failure analysis data. This article examines the key challenges and solutions in creating high-quality TEM samples for advanced semiconductor applications.
Why is TEM sample quality so critical now?
The semiconductor industry faces several technological inflection points requiring superior TEM analysis:
  • Logic devices
    • Gate-all-around (GAA) structures with 2 nm line widths
    • Complex 3D architectures with buried features
    • Increased difficulty in defect detection
  • Memory technologies
    • High-density, high-aspect-ratio channels
    • Shrinking feature sizes
    • Challenging defect visibility
  • Power semiconductors
    • Integration of SiC and GaN substrates
    • Atomic-scale defect detection requirements
    • Complex yield optimization needs
What are the three main challenges in TEM lamella preparation?
1. FIB damage control
  • Amorphous damage increases proportionally with beam voltage
  • Critical threshold: ~1 nm damage per kV
  • Impact: 20 kV process creates 40 nm total damage zone
  • Solution: Low-voltage (500 V) polishing capability
Comparison of ion beam damage at varying voltages (5 kV to 500 V) demonstrating impact on sample quality.
Figure 1: Comparison of ion beam damage at varying voltages (5 kV to 500 V) demonstrating impact on sample quality.
2. Precise cut placement
  • Target accuracy requirements at nanometer scale
  • Impact of misalignment on feature visibility
  • Need for real-time monitoring and adjustment
  • Importance of high-quality SEM imaging
Impact of cut placement accuracy: Off-center vs. centered fin structure in gate.
Figure 2: Impact of cut placement accuracy: Off-center vs. centered fin structure in gate.
3. Lamella thickness control
  • Feature size considerations
  • Sub-10 nm thickness requirements
  • Impact on artifact prevention
  • Relationship to data quality
Thickness effect comparison: 16 nm vs. 7 nm sample showing artifact reduction.
Figure 3: Thickness effect comparison: 16 nm vs. 7 nm sample showing artifact reduction.
How does sample preparation method affect results?
Traditional top-down method:
  • Limitations for sub-30 nm samples
  • Curtaining effects from dissimilar materials
  • Challenges with final polishing
  • Limited success with ultra-thin samples
Top-down preparation limitations showing curtaining effects.
Figure 4: Top-down preparation limitations showing curtaining effects.
Inverted sample preparation:
  • Enables sub-10 nm sample creation
  • Reduces curtaining artifacts
  • Improved structural stability
  • Automated process availability
Best practices for high-quality TEM lamella:
  1. Voltage selection
    • Use lowest practical voltage for final polishing
    • Monitor imaging quality during preparation
    • Adjust parameters based on material properties
  2. Placement control
    • Utilize orthogonal viewing capabilities
    • Implement regular progress checks
    • Maintain precise endpoint control
  3. Thickness management
    • Choose appropriate preparation method based on target thickness
    • Consider structural stability requirements
    • Use wedge milling for ultra-thin samples
Inverted preparation method demonstrating improved sample quality and control and lamella end view for target verification.
Figure 5: Inverted preparation method demonstrating improved sample quality and control and lamella end view for target verification.
Technical specifications and solutions
The Thermo Scientific Helios 6 FIB-SEM offers:
  • Voltage range: 30 kV to 500 V
  • 500 nm resolution at 500 eV
  • Automated sample preparation via Thermo Scientific AutoTEM 6 Software
  • High-precision cut placement capabilities
Things to remember
  1. Sample quality directly impacts data reliability
  2. Sub-10 nm samples require specialized preparation techniques
  3. Advanced automation tools improve success rates
  4. Low-voltage capabilities are essential for damage minimization
Subscribe to Thermo Fisher Scientific's semiconductor updates and resources
Abhi Barve
Written by:
Abhi Barve
Senior Director and General Manager, Semiconductor, Thermo Fisher Scientific
Abhi Barve is Sr. Director and General Manager, Semiconductor with extensive experience in automation, metrology, and advanced semiconductor workflows. His background includes automated transmission electron microscopy, process development analysis, yield learning, and atomic scale characterization.
Read more Barve, Abhi