Harness the power of NGS for your clinical lab with a fast, easy, automated CE-marked IVD system
Designed to make in-house next-generation sequencing (NGS) accessible to clinical laboratories with all levels of sequencing expertise, the Ion Torrent Genexus Dx Integrated Sequencer delivers an effortless user experience while enabling regulatory compliance.
Capabilities
Simply load and go
Place the prepared nucleic acid plate into the Genexus Dx Integrated Sequencer and the instrument will handle the automated library preparation, template preparation, sequencing, and reporting.
Built-in features drive reliable and reproducible results
The on-board sample tracking and barcoding system help you move quickly while reducing errors.
Easily generate custom or pre-packaged reports
Intuitive Genexus Dx Software handles data analysis and reporting, allowing you to interpret results without bioinformatics expertise.
Ordering Information
N° de catalogue Nom Quantité
A53579 -
For In Vitro Diagnostic Use.
Not available in all countries.
CN: 63522