Search
The Ion Torrent Genexus Purification System automates nucleic acid extraction, purification, and quantitation from multiple specimen types on a single platform to provide a consistent and efficient workflow solution for next-generation sequencing (NGS) sample preparation. With just one touchpoint and ten minutes of hands-on time, you can go from sample to purified and quantified nucleic acid in an NGS-ready plate that can be loaded directly onto the Genexus Integrated Sequencer.
The Genexus Purification System can be paired with the Genexus Integrated Sequencer as part of the Genexus System to deliver an end-to-end NGS workflow from biological specimen all the way to the report in a single day with just two user touchpoints and 20 minutes of total hands-on time for the sample preparation, library preparation, sequencing, and data analysis steps combined. This automation helps to reduce user variability and helps ensure tracking of sample information to the final report.
For Research Use Only. Not for use in diagnostic procedures.