Applications of electron backscatter diffraction
EBSD is widely utilized in both industrial and R&D settings because it offers a broad spectrum of detailed information quickly and can characterize large areas. These attributes make it highly appealing across various fields, as it supports quality control (such as defect identification), aids in optimizing production processes, and facilitates failure analysis. Additionally, EBSD plays a crucial role in research and development by providing insights that guide the improvement and development of new materials.
EBSD analysis for metallurgy and materials science
Metallurgy and materials science are the main areas where EBSD brings invaluable insights for optimizing metals processing techniques and for improving products’ lifetimes by better tuning the materials’ properties (strength, ductility, or corrosion resistance). Analyzing the microstructure of metals and alloys can, in fact, support and guide product development and improvement. In addition, metals production companies use EBSD as a form of quality control; variations in the uniformity and consistency of the metals’ microstructures have a direct impact on the performance and reliability of the materials, possibly altering and reducing the quality of the final products.
EBSD manufacturing applications
Many manufacturing industries take advantage of the EBSD capabilities for quality control purposes and root cause analysis of failures. Among others, the automotive, aerospace, and semiconductor industries employ this technique to investigate failures of components and structures, using the results to improve manufacturing processes and future product designs. For example, EBSD is applied in semiconductor manufacturing to analyze crystal defects, grain boundaries, and interfaces within semiconductor materials with the primary goal of increasing the reliability of electronic devices.
EBSD is also crucial for process optimization and for advanced manufacturing. Studying the evolution of a material’s microstructure and crystallographic properties gives important information to optimize processes such as welding, casting, and additive manufacturing.
EBSD analysis for geology and earth sciences research
One of the main capabilities EBSD brings to material characterization is the possibility of identifying phases. For this reason, geologists and earth scientists make use of it for in-depth studies of different mineral phases to understand deformation mechanisms in rocks. EBSD also serves as a tool to understand the crystallographic properties of minerals and rocks and helps in reconstructing geological processes.
EBSD for research and development
Outside of EBSD’s industrial applications, research facilities make extensive use of this technique. Its wealth of information is extremely beneficial for research environments where materials and microstructures are often unknown. Nanotechnology, biology, and biomedical engineering all benefit from the possibility of obtaining insights into the materials’ crystallographic features and interfaces. For example, EBSD is used to analyze the microstructure of biomaterials such as bone implants, dental materials, and tissue scaffolds, with the goal of tuning their mechanical properties.
EBSD finds diverse applications across various industries and research fields, contributing significantly to the understanding, development, and improvement of materials and technologies. Its ability to provide detailed information at the microstructural level makes it a powerful tool for researchers and engineers alike.
Electron backscatter diffraction articles
EBSD Analysis of Metal and Alloy Microstructure
EBSD data processing and results
EBSD analysis strengths and weaknesses
Thermo Scientific Apreo ChemiSEM System
The Apreo ChemiSEM System delivers all-around nanometer or sub-nanometer resolution at an analytical working distance of 10 mm and features fully integrated EDS and EBSD. Its field emission gun (FEG) source makes it possible to analyze a wide range of beam-sensitive materials, composites, magnetic samples, and nano-materials.
For Research Use Only. Not for use in diagnostic procedures.