Search
Western blot analysis of HNRPH3 was performed by loading 35 µg of MiaPaCa-2 lysate in reducing sample buffer and Page Rµler Protein Ladder (Product # 26616) onto a 10% polyacrylamide gel. Proteins were transferred to PVDF membrane using the wet transfer method. Membrane was blocked in Starting Block Blocking Buffer (Product # 37542). HNRPH3 was detected at approximately 37 kDa using a HNRPH3 polyclonal antibody (Product # PA5-41837) at a dilution of 1:1000 in 5% BSA, followed by a goat anti-rabbit IgG DyLight 800 conjµgated secondary antibody (Product # SA5-35571) at a dilution of 1:7000. Data courtesy of Antibody Data Exchange Program..