Search
Optimize your experiments to get the best results. We’ve compiled a detailed knowledgebase of the top tips and tricks to meet your research needs.
View the relevant questions below:
Having problems with your experiment? Visit our
GeneArt Strings DNA Fragments are custom-made, uncloned, double-stranded linear DNA fragments, available in lengths from 150 to 3,000 base pairs (bp), assembled from synthetic oligonucleotides using the same process developed for GeneArt high-quality gene synthesis. GeneArt Strings DNA Fragments can be used to quickly clone your gene of interest. At least 200 ng of GeneArt Strings DNA Fragments are produced within 5 business days (up to 1,000 bp) or 8 business days (from 1,000 to 3,000 bp). GeneArt Strings DNA Fragments can be designed, sequence-optimized, and ordered online and are a cost-effective and smart alternative to clone your expression plasmid.
GeneArt Strings DNA Fragments are PCR amplicons of assembled oligonucleotides; an intermediate product of the gene synthesis production process. This process results in a pool of fragments, and cloning and screening need to be carried out to identify the correct clone. To ensure that correct fragments are present in the pool, GeneArt Strings DNA Fragments are bulk sequence-controlled before shipment.
GeneArt Strings DNA Fragments are shipped dried, ready for resuspension and cloning. Before using, we recommend spinning down the tube contents, adding some water to the bottom of the tube to get the desired concentration, and incubating for at least 1 hour at room temperature (alternatively, incubate at 4 °C overnight). Subsequently, we recommend resuspending the fragments carefully and using immediately. For longer storage, resuspended GeneArt Strings DNA Fragments should be dispensed into aliquots and frozen at –20 °C. Please avoid freeze-thaw cycles.
No. GeneArt Strings DNA Fragments from 150 to 1,000 bp are produced in 5 business days and GeneArt Strings DNA Fragments from 1,000 to 3,000 bp are produced in 8 business days. Depending on the nature of the sequence, production time can vary.
Yes. We recommend using the GeneArt web portal for ordering, where you can use the free gene editing and optimization functions to adjust the sequence to meet your needs. After editing and optimization, please check again that the final sequences of the fragments are exactly as needed for further processing in your lab (e.g., potential homologous overlaps of the fragments or restriction sites and buffer bases needed for cloning).
Yes, we offer GeneArt Strings DNA Libraries from 200 – 2000 bp, containing up to 3 blocks of degenerate nucleotides. Each block can consist of up to 30 bp using the full IUPAC code of DNA nucleotides. Blocks need to be separated by 30 bp of non-degenerate nucleotides.
No, GeneArt Strings DNA Fragments are limited to 150 to 3,000 bp. If your sequence is longer, you need to order it as gene synthesis or assemble your gene from two or more GeneArt Strings DNA Fragments. In addition, theGeneArt Strings DNA Fragments manufacturing process is streamlined to quickly and cost-effectively provide you with your gene product. If your sequence is complex, you may receive a message that it cannot be produced as a GeneArt Strings DNA Fragment due to high complexity. In that case, we recommend modifying your sequence to match the production criteria (given below) or ordering your gene as gene synthesis. In many cases, optimizing your sequence using the portal function enables manufacturing with the GeneArt Strings DNA Fragments process. If it is still not able to be produced, you can manually edit the sequence to enable production.
Important production criteria are:
GeneArt Strings DNA Fragments are PCR amplicons of assembled oligonucleotides, ready for screening to identify the correct clone. GeneArt Strings DNA Fragments are bulk sequence-controlled as part of the QC process to help ensure that your desired sequence is present in the fragment pool.
To identify a correct clone >90% of the time, we recommend using these screening guidelines:
No, subcloning and assembly services are not available for GeneArt Strings DNA Fragments, as they are designed to offer you the fastest and most affordable way to get access to your genes. If you do not want to clone the gene yourself, we recommend that you order full gene synthesis service.
Yes, it is possible to directly assemble two GeneArt Strings DNA Fragments without pre-cloning. We offer several technologies for seamless assembly, e.g., GeneArt Type IIs Assembly or GeneArt Seamless Cloning and Assembly. Depending on the sequence length and the number of subfragments you want to assemble, the screening effort to find a correct clone can be high. Therefore, we generally recommend pre-cloning the subfragments to limit the screening effort required to find a correct clone after assembly.
The answer depends on the specifics of your project. In general, it is advantageous to keep the diversity of a library as low as possible, targeting only the regions of a gene/protein that are likely to be functionally important. The following information can help determine this: crystal structure, conserved motifs, presence of homologs, etc.
Conventional protocols for degenerated library creation (e.g., error-prone PCR) incorporate many unwanted mutations. Moreover, methods like DNA shuffling cannot typically cause recombination of directly adjacent mutations. Synthetic combinatorial libraries, on the other hand, limit the introduction of mutations to defined regions at the precise frequencies requested. In addition, adjacent mutations will be recombined (shuffled) independent of their proximity.
The use of TRIM technology in creating diversity allows for the replacement of complete codons instead of altering single nucleotides. You thus have complete control over which amino acids appear in a given position and in what ratios, while at the same time avoiding the undesired introduction of stop codons. Because complete codons are replaced, TRIM technology also results in fewer out-of-frame mutations than other mutagenesis technologies.
We routinely use optimized codons for the most common expression hosts (human, CHO and E. coli). All other codons are available upon request.
There are a number of reasons:
If you require a degenerate library with an unusual design please send an email to geneartsupport@thermofisher.com. We will consider any project and in the vast majority of cases will find a solution to fulfill your requirements.
Yes, GeneArt Combinatorial Libraries can be randomized using any nucleotide mix you require, down to single digit percentages of given nucleotides (“dirty bottle” approach).
No material is necessary. For library creation, all we need is the sequence file, submitted electronically, and information about the position and nature of the sites you want to randomize. We can provide a quote for your project through our online ordering system. If needed, you can then relate detailed information about your library request to our production scientists prior to starting the project.
Yes. For this purpose, please send in your own customized vector or a commercially available vector.
Two standard options are available:
Three criteria are important for degenerate libraries and are quality-controlled in all GeneArt degenerate libraries (excluding the GeneArt Strings DNA Libraries):
A GeneArt Strings DNA Library is a linear DNA fragment with the following parameters:
Note: GeneArt Strings DNA Libraries are not available with TRIM technology or custom nucleotide mixes. Please inquire about our other library products if you require this type of randomization.
A project can have multiple processes, but can be ordered as a single project. In this way shipping/handling are paid only once.
Yes. We can subclone your gene into a vector you provide. You will receive 5 µg each of your gene in a GeneArt cloning vector and in your vector (plus the quality control information for both constructs).
GeneArt Gene Synthesis Service includes gene optimization (if requested), synthesis of the DNA, cloning into our standard vector, and delivery of 5 µg of lyophilized DNA plus a compact disc containing the in silico analysis and quality control information.
Our proprietary GeneOptimizer software calculates the optimal DNA sequence needed to encode the protein of interest (gene optimization). Adapting the codon usage of the gene to the codon preferences of the expression organism (codon optimization) is just one of the parameters addressed. The GeneOptimizer software currently evaluates up to 50 factors that can compromise mRNA stability, such as extreme GC content, ribosomal binding sites, consensus and cryptic splice sites, repeats, and secondary structures. Increased numbers of stable mRNA molecules often lead to higher yields of protein.
The GeneOptimizer process takes a multi-parameter approach to analyzing the gene sequence. It takes codon usage, GC content, cryptic splice sites, direct repeats, RNA secondary structures, and instability sequences into account when optimizing a gene sequence.
Yes, you can protect specified regions or restriction enzymes sites that you would like to not be optimized by the the GeneOptimizer process. In the portal, under "Optimize sequence", choose "Protect Restriction Sites" and/or "Protect Sequence Parts" from optimization.
Fath et al. (2011) A Standardized Tool to Assess and Enhance Autologous Mammalian Gene Expression.
PLoS ONE 6(3): e17doi:10.1371/journal.pone.0017596
GeneObserver shows the manufacturing status of your web and non-web orders, as well as the completed date so that you can track your gene synthesis project.
GeneArt Strings DNA Fragments are uncloned double-stranded DNA fragments that are limited to length of 3,000 bp. GeneArt Gene Synthesis products are double-stranded DNA fragments that have been cloned into a standard GeneArt vector (pMx). Please note, a sequence may be too complex to be produced as a GeneArt Strings DNA Fragment due to repetitions or high GC content. In this case, our scientists will recommend that you order your gene via gene synthesis. You could try optimizing your sequence through the GeneOptimizer algorithm which, in some cases, could optimize the sequence and allow the DNA Fragments to be ordered.
Production time is based on gene length and choice of shipment speed. Please see this link for the differences between our shipping conditions.
The kit and custom services offer different advantages. Please click here to view a selection guide comparing the technologies, including deliverables and production time, to determine what is best for you.
Genes are typically cloned into a pMx standard vector. These vectors contain:
Please note that standard orders do not guarantee any specific standard vector.
GeneArt Standard vector (pMx) zero cutter restriction enzymes table
We do not offer free choices of restriction sites in our standard production vectors. For our production process, restriction enzymes are added that generally eliminate most of the standard vectors' MCS. The reason the MCS contains more sites is to allow for fallback scenarios in case certain sites cannot be used due to insert sequence intrinsic restrictions.
No, and this cannot be requested. You would need to subclone into a vector of your choice. The only thing you can request as part of the “Geneart vector” is the antibiotic resistance (for a small markup fee).
GeneArt synthesis adds standard restriction sites for automated cloning into GeneArt standard vectors in addition to your selected restriction sites.
Yes, we keep a sample of every GeneArt synthesized product.
In theory, there is no limit. However, very large DNA fragments (greater than 10 kb) need to be cloned into a low-copy vector to ensure genetic stability. Therefore, increasing length leads to increased production time.
You will receive both vectors – your gene in the basic cloning vector, and your gene in the subcloned vector of your choice.
Please provide 5-10µg of vector DNA for your GeneArt custom service. We prefer the DNA dissolved in TE buffer or water, or as a pellet.
There are different compliance regulations/rules for synthesis, including the use of different labs. With Biosafety 2, it takes a longer time to synthesize genes, therefore, these projects may be more expensive. Please note, regardless of your selection, Life Technologies Corporation and/or its affiliates will perform a biosafety evaluation. In the case that you select Biosafety Level 1 but our evaluation requires the DNA to be a Biosafety Level 2, you will be sent a modified quotation according to the Biosafety Level 2 requirements for review.
TSE stands for Transmissible Spongiform Encephalopathy, meaning the media used is soy-based for TSE-free plasmid preps.
You typically would receive the following with your order: vector map, alignment maps, sequencing traces. Extended documentation can include:
You can download your optimized sequence in FASTA format. It can be read in a txt file/format. The FASTA file will give a simple description of the gene, and then give a non-annotated version of the sequence. You can align this file with their original sequence using a free online tools.
Partial deliveries can occur when upon reaching the estimated completion date not all order items have been finalized. Partial shipments of incomplete orders are initiated on a weekly basis. Separate shipping of items does not incur an extra cost.
Generally, we ship immediately upon completion of the project item with the longest estimated turnaround time. In rare cases, completely finalized projects are not shipped until the estimated completion date has been reached. Please keep in mind, if a customer orders gene synthesis and subcloning as a process, they will most likely receive the gene synthesis in a Geneart vector before they receive their gene in their subcloned vector.
No, your custom vector has been banked and sequenced and is available to you in your individual short list “My Portal Vectors” as soon as they have been ordered/used once.
Yes, we will optimize the protein based on the species you choose, to provide you with the optimized gene sequence for your studies.
Yes, all pMx vectors contain an M13 – 20 primer binding site and a M13-R primer binding site. The sequences are as follows:
Single variants and variant bundles (more than one variant process) are variants of a master gene. The variant criteria are as follows:
Variants may contain up to four of the following modifications:
Modifications (whether internal or 5´/3´) must be separated by at least 100 bp.
If a gene falls outside of these criteria, it must be created as a separate gene synthesis instead of as a variant.
Yes. Add “single variant” as a new process, then choose an “upstream service” ("gene synthesis", "already at Geneart", or "I will provide this master gene"). If you are providing your own gene, you will need to fill in the field for the complete sequence of the vector + insert. If it is a plasmid only, you will enter the mutagenized area as the “insert” and the rest of the plasmid as the “vector”. The “insert” can be up to 4 kb long.
Using an alternative production procedure with pre-prepared expression vectors, our GeneArt custom service team can clone your synthesized genes directly into selected Life Technologies expression plasmids, saving 4-5 business days compared to the standard workflow with cloning into a pMx series vector first. Please visit this page for the differences between Express cloning and Classical cloning.
You should receive your synthesized gene in the selected expression vector. Please note, no additional copy in a pMx cloning vector will be provided as with traditional GeneArt Cloning Services.
The following vectors are currently available:
Genes qualifying for Express cloning must be <4 kb and not complex (optimization of your sequence can reduce complexity).
Please use the following link to explore our synthetic biology services which include: vector construction, cDNA library construction, protein expression services, directed evolution services, plasmid services, and cell line & protein services.
TALs are transcription activator-like effector proteins that are naturally occurring transcriptional activators. GeneArt Precision TALs allow researchers to determine the exact DNA sequence they would like to have their functionality delivered to and have specific TAL genes built to perform the function.
GeneArt Site-Directed Mutagenesis Kits
The kits are almost the same, but the PLUS kit contains an improved 2X GeneArt Enzyme mix with protocols optimized for multi-site mutagenesis (up to 3 sites in plasmids up to 14kb). The PLUS kit can also be used for single-site mutations of up to 25 nucleotides, while the original kit can be used for single substitution/deletion/insertions up to 12 nucleotides in plasmids up to 14kb. A web tool is available for primer design, that can also be used for the original kit.
We do not offer the DNA methylase from GeneArt Site-Directed Mutagenesis kits as a separate product.
The concentration is 32 mM.
The primer should be 30-45 nucleotides in length with the mutation centrally located. The mutation should be flanked by at least 15-20 nucleotides on either side, depending on the size of the mutation, with the primer length no longer than 45 nucleotides. We recommend using our online GeneArt primer design tool.
During the first cycle of PCR on the circular template, the polymerase will generate two linear products that are able to anneal and undergo primer extension in the next cycle (see diagram below). PCR products that incorporate the mutation at both ends will accumulate in subsequent cycles. After PCR, the recombination reaction results in circularization of the PCR product which enhances the mutagenesis efficiency.
In general, the annealing temperature used will be between 55-64 °C, which is what we recommend for PCR with AccuPrime Pfx. The Tm of the primers will be about 5°C above the annealing temperature. For example, the Tm of the top strand control primer that comes with the kit is 65 °C (nearest neighbor method).
We recommend shortening the primer length for GC-rich sequences, since a very high Tm may result in nonspecific bands. A previous customer reported having success using 21-nt primers with a Tm of ~80 °C.
The polymerase should not have a problem reading through AT-rich regions. Only GC-rich regions can be difficult for polymerases because of the higher Tm.
The primer design guidelines in the GeneTailor kit (discontinued) should work as long as the primers have an overlapping region of 15-20 nucleotides. However, to make primer design straightforward we recommend that users switch and use the GeneArt guidelines only.
This is most likely due to rearrangement caused by hairpin formation of the primer during PCR. You can try and increase the annealing temperature of the PCR step to relax secondary structure formation. Screening more colonies should also help.
In general, the PCR enhancer increases the yield of PCR product. It has nothing to do with DNA methylation. The 10x enhancer is optimized for Accuprime Pfx and it may not work with other polymerases. If you need to optimize the PCR (for annealing temp, extension time, etc.) you should do standard optimization with AccuPrime Pfx but without the enhancer.
We currently do not supply this kit with a DNA polymerase. We do recommend purchasing AccuPrime Pfx DNA polymerase for your site-directed mutagenesis experiments with this kit, as we have optimized the conditions and protocol using this enzyme.
This is most likely due to rearrangement caused by hairpin formation of the primer during PCR. You can try and increase the annealing temperature of the PCR step to relax secondary structure formation. Screening more colonies should also help.
For GeneArt mutagenesis services, we will need the DNA or amino acid sequence of the gene you would like to mutagenize, the host organism you plan to use (this is important for gene optimization if you choose to include it with your request), the restriction sites you need at the 5’/3’ ends and/or to avoid internally, and whether or not you want any other added motifs (e.g., Kozak sequence, stop codons, etc.).
Variants may contain up to four of the following modifications:
Note: Modifications (whether internal or 5´/3´) have to be separated by at least 100 bp
For Research Use Only. Not for use in diagnostic procedures.