Search
Blood Plasma and Serum Preparation |
Separating serum and plasma from blood involves distinct processes. Serum is the liquid fraction of whole blood collected after allowing the blood to clot. The clot is removed through centrifugation, and the resulting supernatant, designated as serum, is carefully extracted using a Pasteur pipette. Plasma is produced when whole blood is collected in tubes treated with an anticoagulant. In these tubes, the blood does not clot. The cells are removed by centrifugation, and the supernatant, designated as plasma, is carefully extracted from the cell pellet using a Pasteur pipette. These methods are crucial for various diagnostic and research applications.
Search available antibody pairs and ELISA kits Browse available ELISA reagents
| The commercially available serum tubes are as follows: | |
|---|---|
| Red | No anticoagulant. |
| Red with black | Treated with gel to help to separate the clot. |
|
|
|
|
|
Note:Samples which are hemolyzed, icteric, or lipemic can invalidate certain tests.
| The commercially available plasma tubes are as follows: | |
|---|---|
| Lavender | Treated with EDTA. |
| Blue | Treated with citrate. |
| Green | Treated with heparin. |
| Grey | Treated with potassium oxalate/sodium fluoride. |
| Yellow | Treated with potassium oxalate/sodium fluoride. |
|
|
|
|
Note: Hemolyzed, icteric, or lipemic samples can invalidate certain tests.