Vanquish Horizon UHPLC System
The Thermo Scientific Vanquish Horizon UHPLC system provides unrivaled performance and throughput for applications requiring high-end UHPLC, ideal for research and development labs. With a fully biocompatible flowpath and ultra-low gradient delay volume, it offers superior separations with unsurpassed retention time and peak area precision, excellent detector sensitivity and lower baseline noise for the most demanding LC and LC-MS workflows. Like all members of our Vanquish product line, Vanquish Horizon UHPLC systems offer hardware precision, detector sensitivity, and simplicity of operation.
Build your ideal HPLC or UHPLC with our step-by-step configurator
Highlights
Powerful separations
Vanquish Binary Pump H delivers industry-leading UHPLC flow performance using SmartFlow, SmartStroke and Adaptive Thermal Effect Compensation (ATEC) technologies at maximum operating pressures of up to 1500 bar (22,000 psi). 
Mastering most challenging LC-MS applications
Industry-leading performance results in the highest retention time stability, chromatographic resolution and selectivity. This supports LC-MS workflows using ultra-fast gradients and produces the highest quality data with respect to precision, accuracy and reproducibility.
Automated sample handling
Vanquish Split Sampler HT offers outstanding speed and injection precision using SmartInject technology. It has a default vial capacity of 216 samples, which can be extended up to 8832 samples with the Vanquish Charger sampler extension. 
Sensitive analyte detection
The Vanquish Diode Array Detector HL includes LightPipe technology, featuring a long light path with low dispersion for fast and sensitive detection. We offer additional detection options, including near-universal charged aerosol detection, fluorescence detection and mass spectrometric detection. 
Biocompatible, easy and reliable tool-free connections
The fluidic pathway of the Vanquish Horizon UHPLC system is fully biocompatible and fitted with Thermo Scientific Viper fingertight fittings, which support near-zero dead volume connections and enable sharper peaks and tool-free setup.
CDS software
The Vanquish Horizon UHPLC system integrates with Thermo Scientific Chromeleon Chromatography Data System (CDS) with compliance tools, networking capabilities, automation, and data processing. We offer standard instrument integration (SII) packages to connect Thermo Scientific HPLC systems to other CDS and MS software programs.
Featuring the ACT Ecolabel
Thermo Scientific Vanquish Horizon UHPLC systems are certified with the prestigious My Green Lab® ACT® Ecolabel awarded by My Green Lab, Inc. The My Green Lab ACT Ecolabel serves as the premier ecolabel for lab products, encompassing consumables, chemicals, and equipment. Standing for Accountability, Consistency, and Transparency (ACT), this certification offers clear, standardized information about a product’s environmental impact across its life cycle covering manufacturing, energy and water use, packaging, and end-of-life disposal. Leverage the My Green Lab ACT Ecolabel to make informed purchasing choices based on transparent environmental data so you can advance sustainability without sacrificing performance.
Videos
System specifications
The table below provides an overview of the Vanquish Horizon UHPLC system specifications for a standard setup. Not all specifications are listed. Your specifications may differ based on the components of your system. Find the specification sheets for the system components in the Component specifications, below.
Category  Vanquish Horizon UHPLC System
System pressure, maximum 1500 bar
Flow range 0–5 mL/min
Pump options (default gradient delay volume) Binary (35 µL)
Biocompatible Yes
pH range  2–12, chloride concentration up to 1 mol/L
System design Modular
Touchscreen user interface display compatible (optional) No
Solvent monitoring system compatible (optional) Yes
Customizable gradient delay volume to simplify method transfer  Yes, with metering device
SmartInject technology for maximum injection precision and improved column lifetime
Yes
Injection volume range Standard: 0.01-25 µL
Optional: 0.01-100 µL
Sample cooling
Yes, 4-40°C
Sample capacity (standard)
Based on standard 12 mm OD vials (≤ 1.5 mL)
4 racks of 54 vials  (≤ 1.5 mL) each, other rack formats available
+ capacity of 12 × 22.5 mm OD vials (≤ 10 mL) in the carousel
Sampler extension available
Yes, with Vanquish Charger
Column compartment thermostatting Yes, 5-120 °C 
Still air and forced air 
Column compartment pre-heater Active and passive
Column compartment post-column cooler Yes
Detector choices
Vanquish Charged Aerosol Detector
Vanquish Diode Array Detector
Vanquish Fluorescence Detector
Vanquish Multiple Wavelength Detector
Vanquish Variable Wavelength Detector
Mass Spectrometer
Component specifications
Standard Vanquish Horizon UHPLC systems are made up of the following components. Talk to your local sales rep to discuss your application needs. Download the associated specifications sheet for more detail on each component.
  Vanquish Horizon UHPLC
Pump
Download Specification Sheet
Vanquish Binary Pump H
Autosampler
Download Specification Sheet
Vanquish Split Sampler HT
Extended sample capacity (optional)
Download Specification Sheet
Vanquish Charger (optional)
Column compartment
Download Specification Sheet
Vanquish Column Compartment H
Virtually zero dead volume system connections
Download Specification Sheet
Viper UHPLC Fittings Kits
Detectors
Visit our HPLC and UHPLC Detectors page
Optional components: Fraction Collector Visit our Vanquish Fraction Collector FT page
Contact us to get a quote or discuss an HPLC/UHPLC solution
Our general purpose product lines, including LC-MS products, are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks. They are designated for Research Use Only Applications.
For Research Use Only - Not For Use In Diagnostic Procedures.