Analytical techniques for automotive materials characterization
Analyzing advanced automotive materials and components requires techniques that deliver precise chemical, structural, and surface-level insight across a wide range of materials and manufacturing processes. Thermo Scientific instruments support key automotive analytical needs through the following core techniques.
Elemental and phase analysis
Accurate elemental identification and quantification are essential for alloy development, impurity detection, and contamination control. Elemental analysis supports materials qualification, quality control, and failure investigations by revealing elemental composition, distribution, and phase chemistry across metals, coatings, polymers, and battery materials.
Layered filler. From left: BSE image, selected elements, and quantitative elemental image.
Nanoparticle analysis
Understanding particles, inclusions, and nanoscale features is critical for controlling material performance and reliability. High-resolution imaging and automated particle analysis help detect, classify, and statistically evaluate nanoparticles and inclusions, helping manufacturers better understand the evolution of microstructural defects that influence mechanical strength, wear, and fatigue behavior.
Wide-area automated TEM image with color-coded energy dispersive X-ray spectroscopy (EDS) elemental maps for particle analysis and sizing.
Surface and interface analysis
Many automotive failures and performance limitations originate at surfaces and interfaces. Surface-sensitive techniques provide insight into coatings, passivation layers, corrosion products, and chemical states. These analyses are essential for evaluating surface treatments, adhesion quality, corrosion resistance, and paint or coating defects.
Sputter depth profiles of unpassivated and passivated steel surfaces.
Sample preparation
High-quality analysis begins with proper sample preparation. Advanced preparation techniques help enable precise cross-sectioning, site-specific access, and damage-free exposure of regions of interest across complex automotive materials such as multilayer coatings, additive-manufactured parts, and battery components.
SEM backscattered electron (BSE) image of an auto body coating cross section.
ToF-SIMS surface chemical analysis
Time-of-flight secondary ion mass spectrometry (ToF-SIMS) provides highly surface-sensitive chemical analysis and imaging. It helps manufacturers detect trace-level organic and inorganic species, map the chemical composition of thin films and interfaces, and perform depth profiling of multilayer structures. This technique is valuable for investigating coatings, adhesives, battery interfaces, and surface contamination in automotive materials.
Electron backscatter diffraction (EBSD) image of a weld, HAZ, and base metal regions. Courtesy of WMG (Warwick Manufacturing Group), University of Warwick.
Crystallographic and phase analysis
Grain structure, phase distribution, and crystallographic orientation strongly influence mechanical and thermal properties. Crystallographic analysis supports process optimization, development of heat treatments, and root-cause failure analysis by linking microstructure to performance.
EBSD map (IPF Z) of an aluminum alloy (AA2024).
Particle, inclusion, and technical cleanliness analysis
Automated particle and inclusion analysis helps manufacturers detect, classify, and statistically evaluate contaminants, wear particles, and non-metallic inclusions. These workflows support technical cleanliness requirements by measuring particle size, shape, and composition, and by generating standards-based reports for automotive quality control. High-throughput analysis helps improve process monitoring, reduce variability, and support consistent compliance across manufacturing and production environments.
SEM image of an aluminum matrix showing microcracks. Sample courtesy of the University of Science and Technology Beijing.
Digital analysis of morphology and chemistry with AI
Thermo Scientific Avizo Software combines advanced electron microscopy (EM) visualization, quantification, and AI-driven image analysis to accelerate automotive materials characterization. With Avizo Software, scientists and engineers can segment and analyze complex microstructures in battery materials, lightweight alloys, composites, catalysts, and electronic components. AI-assisted segmentation helps to automatically identify phases, particles, pores, defects, and interfaces, significantly reducing analysis time while providing a repeatable, quantified 
Analyzing coating performance with a Thermo Scientific Helios FIB-SEM and Avizo Software’s AI-accelerated analysis.
For Research Use Only. Not for use in diagnostic procedures.