Search
The E-Gel® agarose gel electrophoresis system is a complete bufferless system for agarose gel electrophoresis of DNA samples.
The major components of the system are:
E-Gel® pre-cast agarose gels are self-contained gels that include electrodes packaged inside a dry, disposable, UV-transparent cassette. The E-Gel® agarose gels run in a specially designed device that is a base and power supply combined into one device.
Advantages of E-Gel®
Using E-Gel® agarose gels for electrophoresis of DNA samples offer the following advantages:
Medium/High-Throughput E-Gel® Electrophoresis System
The Medium/High-Throughput E-Gel® Electrophoresis System is designed for electrophoresis of 48-96 DNA samples per gel. This system is compatible for use with multichannel pipettors or automated liquid handling systems. For details on this system, see below.
The system consists of the following components:
E-Gel® 48 gels
Each E-Gel® 48 gel contains 48 sample lanes and 4 marker lanes and is designed for medium-throughput agarose electrophoresis of nucleic acids.
E-Gel® 96 gels
Each E-Gel® 96 gel contains 96 sample lanes and 8 marker lanes in a patented, staggered-well format and is designed for high-throughput agarose electrophoresis of nucleic acids.
E-Base™ Electrophoresis Device
The E-Base™ is a base and a power supply all in one device and is an easy-to-use, pre-programmed device specifically designed for electrophoresis of E-Gel® 48 and 96 gels.
E-Holder™ Platform
The E-Holder™ Platform is designed to hold E-Gel® 96 gels during robotic loading. The E-Holder™ is used to load multiple gels on a robotic platform while other gels are running on the E-Base™.
E-Editor™ 2.02 Software
The E-Editor™ 2.02 software allows you to quickly reconfigure digital images of E-Gel® 48 or 96 gel results for analysis and documentation. The E-Editor™ 2.02 software can be downloaded for free.
Choosing a Gel for Your Application
To obtain the best results for your application, it is important to choose the correct agarose percentage and well format.
The table below lists the various types of gel and resolution for each gel type.
Gel Type | No. Rows | No. Wells | Sample Volume | Run Length | Run Time | % Agarose | Resolution |
E-Gel
®48 Gel
|
2*
|
48 sample
4 marker
|
10-15 µl
15 µl
|
3.2 cm
|
20 min.
|
1%
2%
4%
|
400 bp-10 kb
50 bp-3 kb
10 bp-400 bp
|
E-Gel
®96 Gel
|
8*
|
96 sample
8 marker
|
10-20 µl
20 µl
|
1.6 cm
|
12 min.
|
1%
2%
|
1 kb-10 kb
100 bp-2 kb
|
*Wells compatible for loading with a multichannel pipettor.
System Components
The Medium/High-Throughput E-Gel® Electrophoresis System is compatible for use with multichannel pipettors or automated liquid handling systems.
The system consists of the following components:
Applications
E-Gel® 48 and 96 agarose gels are suitable for analyzing multiple samples:
E-Gel® 48 Gels
E-Gel® 48 gels are self-contained, pre-cast agarose gels that include agarose, a proprietary buffer system, ethidium bromide, and electrodes packaged inside a dry, disposable, UV-transparent cassette. Each E-Gel® 48 gel contains 48 sample lanes and 4 marker lanes and is designed for medium-throughput agarose electrophoresis of nucleic acids. This configuration provides a 3.2 cm run length.
The 4% E-Gel® 48 gels are prepared with high-resolution agarose to ensure quality resolution of DNA fragments below 400 bp. The wells of the E-Gel® 48 gel are compatible for loading with a multichannel pipettor. The lane numbers are labeled with fluorescent dye that transfers to the image and allows tracking of your samples during photo documentation of the gel.
In addition, each E-Gel® 48 cassette is labeled with an individual barcode to facilitate identification of the gel using commercial barcode readers.
Separation Range for E-Gel® 48 Gels
The separation range for E-Gel®48 gels is listed below:
Sample Range | bp Separation |
1% E-Gel®48 | |
400 bp-600 bp
|
50 bp
|
600 bp-1 kb
|
100 bp
|
1 kb-4 kb
|
500 bp
|
4 kb-10 kb
|
1 kb
|
2% E-Gel®48 | |
100 bp-300 bp
|
25 bp
|
300 bp–700 bp
|
50 bp
|
700 bp-1200 bp
|
100 bp
|
1200 bp-2000 bp
|
200 bp
|
4% E-Gel®48 | |
5 bp-40 bp
|
5 bp
|
40 bp-80 bp
|
10 bp
|
80 bp-175 bp
|
20 bp
|
175 bp-300 bp
|
50 bp
|
300 bp-600 bp
|
100 bp
|
E-Gel® 96 Gels
E-Gel® 96 gels are self-contained, pre-cast agarose gels that include agarose, a proprietary buffer system, ethidium bromide, and electrodes packaged inside a dry, disposable, UV-transparent cassette. Each E-Gel® 96 gel contains 96 sample lanes and 8 marker lanes in a patented, staggered-well format. The wells of the E-Gel® 96 gel are compatible with the standard 96-well plate format for automated loading.
In addition, each E-Gel® 96 cassette is labeled with an individual barcode to facilitate identification of the gel using commercial barcode readers. The lane numbers are labeled with fluorescent dye that transfers to the image and allows tracking of your samples during photo documentation of the gel.
During electrophoresis, samples migrate between the wells of the row below. For example, the bands of the lane B11 migrate between well C11 and C12. This configuration provides a 1.6 cm run length, allowing resolution between 100 bp and 10 kb. The staggered well format of the gel cassette is compatible with automated liquid handling devices that use 8-, 12-, or 96-tip loaders. During sample loading, the samples will fall onto the slopes of the wells and be drawn into the wells by capillary force.
Diagram of E-Gel® 96 Cassette
A diagram of the E-Gel® 96 cassette is shown below.
E-Base™
Two types of bases are available from Invitrogen:
The Mother E-Base™ (Catalog no. EB-M03) has an electrical plug that can be connected directly to an electrical outlet and is used for electrophoresis of one E-Gel® 48, E-Gel® 96, or E-PAGE™ 96 gels available from Invitrogen.
The Daughter E-Base™ (Catalog no. EB-D03) connects to the Mother E-Base™, and together they can be used for the electrophoresis of two or more, E-Gel® 48, E-Gel® 96, or E-PAGE™ 96 gels available from Invitrogen.
Note: The Daughter E-Base™ does not have an electrical plug and cannot be used without a Mother E-Base™.
Mother E-Base™
Each Mother E-Base™ has a pwr/prg (power/program) button (right side) and a time button (left side) on the lower right side of the base. The lower left side of each Mother E-Base™ contains a light LED and a digital display. The gel cassette is inserted into the two electrode connections. The Mother E-Base™ is connected to an electrical outlet with the electrical plug.
The E-Base™ is pre-programmed with 2 programs specific for each gel type as described below:
| Program | Gel Type | Run Parameters |
|---|---|---|
| EG | E-Gel® 96 | Time: 12 minutes |
Mother E-Base™
Daughter E-Base™
The Daughter E-Base™ is similar to the Mother E-Base™ except the Daughter E-Base™ does not have an electrical cord and cannot be connected to an electrical outlet.
The Daughter E-Base™ is connected to a Mother E-Base™ or to another Daughter E-Base™ (already connected to a Mother E-Base™). Once connected to a Mother E-Base™, each Daughter E-Base™ is designed to function independently of the Mother E-Base™ or other Daughter E-Bases™.
Mother E-Base™/Daughter E-Base™ 
E-Holder™ Platform
The E-Holder™ Platform is designed to hold E-Gel® 96 gels during robotic loading. Use the E-Holder™ when you need to load multiple gels on a robotic platform while the other gels are running on the E-Base™.
Note: The E-Holder™ is not a power supply unit, cannot be connected to an electrical outlet, and cannot be used to run gels.
E-Editor™ 2.02 Software
The E-Editor™ 2.02 software allows you to quickly reconfigure digital images of E-Gel® 48 or 96 gel results for analysis and documentation.
Capture an image of the gel and then, use the E-Editor™ 2.02 software to:
Align and arrange the lanes in the image
Save the reconfigured image for further analysis
Copy and paste selected lanes or the entire image into other applications for printing, saving, e-mailing, and/or publishing on the Web.
E-Editor™ 2.02 software can be downloaded for free from our Web site. Follow the instructions to download the software and user manual.
Product Specifications
E-Gel® 48 Gel Specifications
Each E-Gel® 48 gel contains 48 sample wells and 4 marker wells (M).
Cassette Size
|
13.5 cm (l) x
10.8 cm (w) x0.67 cm (thick)
|
Gel Thickness
|
3.7 mm
|
Gel Volume
|
50 ml
|
Gel Percentage
|
1%, 2%, and 4%
|
Well Depth
|
3 mm
|
Dimensions of the Well
|
3.6 mm (l) x 2.2 mm (w)
|
Running Distance
(one well to the next)
|
32 mm
|
Space between Well Center
|
Introduction
For optimal results, follow the guidelines for preparing your DNA sample as described in this section.
Materials Needed
Amount of DNA
Use 20-100 ng DNA per band for samples containing one unique band, or up to 500 ng per lane for samples containing multiple bands. If you are unsure how much to use, test a range of concentrations to determine the optimal concentration for your particular sample. Excess DNA will cause poor resolution.
Loading Method
The DNA samples are loaded in E-Gel® agarose gels using the One-Step Loading or Two-Step Loading method.
The One-Step Loading method is routinely used to load E-Gel® agarose gels. The Two-Step Loading method is used to load E-Gel® agarose gels, if the One-Step Loading method produces fuzzy or indistinct bands, or the gel was removed from its plastic pouch for more than 30 minutes. Prepare loading buffer and sample volumes based on the loading method as described below.
Total Sample Volume
The recommended total sample volume for each gel type is listed in the table below.
Note: For best results, keep all sample volumes uniform. If you do not have enough samples to load all wells of the gel, load an equal volume of buffer containing the same salt concentration as samples into any empty wells.
Gel Type | One-Step Loading | Two-Step Loading |
E-Gel
®48 gel
|
15 µl
|
10 µl
|
E-Gel
®96 gel
|
20 µl
|
10 µl
|
Loading Buffer
Loading buffer is optional for One-Step Loading method but is required for the Two-Step Loading method for E-Gels
You can load your samples directly into the wells, if no loading buffer is used. If you are using loading buffer, mix the required amount of DNA with the loading buffer (see preparing samples).
We recommend using a loading buffer with the following formulation:
For One-Step Loading method, use a loading buffer that yields a final concentration of 10 mM Tris-HCl, pH 7.5; 1 mM EDTA; 0.005% bromophenol blue; 0.005% xylene cyanol FF.
For Two-Step Loading method, use a loading buffer that yields a final concentration of 10% glycerol (or 6% Ficoll 400) 10 mM Tris-HCl, pH 7.5; 1 mM EDTA; 0.005% bromophenol blue; 0.005% xylene cyanol FF
If you are using 10X BlueJuice™ Gel Loading Buffer or TrackIt™ Loading Buffer available from Invitrogen, dilute this buffer 50-to 200-fold to obtain optimal results with E-Gel® agarose gels. For Two-Step loading with 10X BlueJuice™ Gel Loading Buffer or TrackIt™ Loading Buffer dilute this buffer 50- to 200-fold and add 10% glycerol.
High Salt Samples
Important: To obtain the best results, prepare your high salt samples as described below.
Samples containing > 50 mM NaCl, 100 mM KCl, 10 mM acetate ions, or 10 mM EDTA (i.e. certain restriction enzyme and PCR buffers) will cause loss of resolution on E-Gel® agarose gels. Dilute samples 2- to 20-fold as described:
Gel Type | One-Step Loading | Two-Step Loading |
E-Gel
®48 gel
|
15 µl
|
10 µl
|
E-Gel
®96 gel
|
20 µl
|
10 µl
|
Preparing Samples
Prepare your samples based on the loading method used as described below:
One-Step Loading | Two-Step Loading | |
Total Sample Volume
| E-Gel®48 gel: 15 µl E-Gel®96 gel: 20 µl
Add deionized water to the required amount of DNA to bring the total sample volume to that listed above.
| E-Gel®48 gel: 10 µl E-Gel®96 gel: 10 µl
Mix the required amount of DNA with a glycerol loading buffer (see below) to obtain a final glycerol concentration of 10% in a total sample volume of 10 µl.
When loading into the gel, first load 5 µl (E-Gel
®48 gel) or 10 µl (E-Gel
®96 gel) of deionized water directly into each well and
then add 10 µl of sample containing 10% glycerol.
|
Loading Buffer
| Optional
Instead of water, you may use a loading buffer that yields a final concentration of 10 mM Tris-HCl; 1 mM EDTA, pH 7.5; 0.005% bromophenol blue; and 0.005% xylene cyanol FF.
To use 10X BlueJuice
™ TrackIt™ Loading Buffer, dilute this buffer 50- to 200-fold to obtain the optimal dye concentration. Gel Loading Buffer or or TrackIt
™ Loading Buffer, dilute this buffer 50- to 200-fold to obtain the optimal dye concentration.
|
Prepare a loading buffer that yields a final concentration of
10% glycerol (or 6% Ficoll 400), 10 mM Tris-HCl, pH 8.1; 1 mM EDTA, pH 7.5; 0.005% bromophenol blue; 0.005% xylene cyanol FF.
To use 10X BlueJuice™TrackIt™ Loading Buffer, dilute this buffer 50- to 200-fold and then add glycerol to a final concentration of 10% in the sample. Gel Loading Buffer or or TrackIt
™ Loading Buffer, dilute this buffer 50- to 200-fold to obtain the optimal dye concentration.
|
DNA Molecular Weight Markers
We recommend using the following DNA molecular weight markers for different types of E-Gel® agarose gels to obtain good resolution.
Note: Supercoiled DNA molecular weight markers may produce a slightly fuzzy pattern when run on E-Gel® agarose gels containing ethidium bromide. Consider using Clear E-Gel® agarose gels and post-staining with ethidium bromide.
Product | Markers | Catalog no. | Amount Used |
E-Gel®48 gels | |||
1%
|
1 Kb Plus DNA Ladder
|
10787-018
|
Load 100-250 ng of markers in a volume of 15 µl in marker well. Use a buffer containing the same salt concentration as your samples.
|
TrackIt
™ 1 Kb Plus DNA Ladder
|
10488-085
| ||
500 bp DNA Ladder
|
10594-018
| ||
E-Gel
® High Range DNA Marker
|
12352-019
| ||
2%
|
TrackIt
™ 50 bp DNA Ladder
|
10488-043
| |
TrackIt
™ 100 bp DNA Ladder
|
10488-058
| ||
50 bp DNA Ladder
|
10416-014
| ||
100 bp DNA Ladder
|
15628-019
| ||
Low DNA Mass Ladder
|
10068-013
| ||
E-Gel
® Low Range Quantitative DNA Marker
|
12373-031
| ||
4%
|
TrackIt
™ 25 bp DNA Ladder
|
10488-022
| |
TrackIt
™ 50 bp DNA Ladder
|
10488-043
| ||
25 bp DNA Ladder
|
10597-011
| ||
50 bp DNA Ladder
|
10416-014
| ||
E-Gel
® Low Range Quantitative DNA Ladder
|
12373-031
| ||
E-Gel®96 gels | |||
1%
|
E-Gel
®96 High Range DNA Marker
|
12352-019
|
Load 100-250 ng of markers in a volume of 20 µl in marker well. Use a buffer containing the same salt concentration as your samples.
|
2%
|
E-Gel
® Low Range Quantitative DNA Ladder
|
12373-031
| |
Introduction
This section describes the procedure for loading and running samples on E-Gel® 48 and E-Gel® 96 gels using the Mother E-Base™ and Daughter E-Base™.
The Mother E-Base™ and Daughter E-Base™ are designed to fit most robotic platforms allowing you to load and run E-Gel® 48 and 96 Gels directly on the robot.
If you need to load multiple gels on a robotic platform while other gels are running on the E-Base™, use an E-Holder™ Platform.
The run time for the E-Gel® 96 gel is 12 minutes and E-Gel® 48 gels is 20 minutes.
The E-Gel® 48 and 96 gels can only be used once. Do not re-use.
The E-Gel® 48 and 96 gels are compatible with the E-Gel® 96 mother base and daughter base available previously from Invitrogen. For instructions on using the gels with E-Gel® 96 mother base and daughter base, see Running E-Gel® 48/96 Gels.
Using the Barcode
Each E-Gel® 48 and 96 gel is labeled with an individual barcode (with a number). The barcode facilitates identification of each gel cassette during the electrophoresis of multiple gels. Each E-Gel®48 and 96 gel contains an EAN 39 type of barcode, which is recognized by the majority of commercially available robotic barcode readers. Refer to the manufacturer’s instructions to set up the barcode reader.
Note: When capturing an image of the E-Gel® 48 or 96 gel, note that the barcode label is easily overexposed. To ensure that the barcode label is distinct and readable in the image, experiment with different shutter settings for your particular camera.
Aligning the Robotic Loading Assembly
The wells of the E-Gel® 96 gel are staggered to provide maximum run length (see Figure 1, below). For proper loading of samples, it is important to program your robotic loading system to set the A1 tip of the 8, 12 or 96-tip robotic head over the E-Gel® 96 gel cassette as described below.
Set the position of the first tip, approximately 1 mm above the slope of the A1 well (see Figure 2, below). This will ensure that the remaining tips are aligned above the slopes of the remaining wells. Refer to the manufacturer’s manual of your robot to program this setting. After programming the setting, load your samples. During loading, the samples will fall onto the slopes of the wells and be drawn into the wells by capillary force. 
For optimal results, load each E-Gel® 48 and 96 gel within 30 minutes of removing the gel from the plastic pouch and run the gel within 15 minutes of loading. If a gel has been out of its plastic pouch for more than 30 minutes, you must use the Two-Step Loading method described.
Do not pre-run E-Gel® 48 and 96 gels.
Store and run E-Gel® agarose gels at room temperature.
Selecting Program on E-Base™
The recommended program for E-Gel® is EG, and the run time for E-Gel® 48 gels is 20 minutes and E-Gel® 96 gels is 12 minutes.
You will need to select an appropriate program on the base prior to inserting a gel into the base. Note: If you had previously set the E-Base™ to the desired program or set the time, the last used program or time is displayed.
| Gel Type | Program |
| E-Gel® 96 | EG |
| E-Gel® 48 | EG and then manually change time to 20 minutes by pressing and holding the time button until 2 minutes is displayed. |
Setting the Time
The initial default time setting on an E-Base™ for program EG is 12 minutes. Follow instructions below to increase or decrease the time setting, if desired.
Do not run an E-Gel® 96 gel for more than 20 minutes or E-Gel® 48 gel for more than 30 minutes.
To increase or decrease the default run time when no cassette is inserted on the base, use the following steps:
If the time button is not released, the time setting will increase until it reaches 00. To begin cycling through the numbers again, starting from 00, press the time button again.
Note: To increase the run time when a cassette is inserted, press and release the time button to increase the time setting by 1-minute intervals or press and hold the time button to increase the time continuously.
To increase the run time while a run is in progress, see below. To manually interrupt or stop a run, see Running E-Gel® 48/96 Gels.
Inserting Gel in the E-Base™
Each E-Gel® 48 and 96 gel is supplied individually wrapped and ready for use. Use short, rigid tips for robotic loading.
To load your samples on the E-Holder™.
Note: If you accidentally inserted a gel into the base before selecting program EG, remove the gel, select program EG, and then reinsert the gel in the base.
Method of Loading Samples
We recommend the following methods of sample loading based on the gel type:
| GelType | Method of Loading |
| E-Gel® 48 | Manual, multichannel pipettor (load samples into alternate wells of the gel followed by a second round of loading into the remaining wells), or robotic loading devices (8- or 12-tip) |
| E-Gel® 96 | Manual, multichannel pipettor, or robotic loading devices (8-, 12-, or 96-tip) |
Two-Step Loading Method
First load deionized water into each well (including sample and empty wells). Then load prepared sample into each sample well. Do not premix. Load sample buffer containing the same salt concentration as your sample into any remaining empty wells.
Load the appropriate DNA markers in the marker (M) wells of an E-Gel® 48 and 96 gels
Using E-Base™
Instructions for running E-Gel® 48 and E-Gel® 96 gels in a Mother E-Base™ or Daughter E-Base™ are provided below.
Note: It is not necessary to have a gel in the Mother E-Base™ if you are using a Daughter E-Base™. However, the Mother E-Base™ must be plugged into an electrical outlet.
Note: The bands in the gel will diffuse within 20-40 minutes.
We recommend that you disconnect the Mother E-Base™ from the electrical outlet when not in use for a prolonged period of time.
Interrupting an Electrophoresis Run
You can interrupt an electrophoresis run at any time by pressing and releasing the pwr/prg button to stop the current. The stopped current is indicated by a steady red light and the digital display will flash to indicate that the run was interrupted. You can remove the gel from the E-Base™ to check the progress of the run. Then:
In case of an external power failure (loss of electricity or the electrical cord is accidentally removed from the outlet), the run will continue when the power resumes. The Mother E-Base™ or Daughter E-Base™ will signal the end of the run as described above, except the light will be an alternating red/green to indicate that an external power failure had occurred during the run.
Maintaining E-Base™
The surfaces of the Mother E-Base™ and Daughter E-Base™ should be kept free of contaminants. To clean, disconnect bases from power source and wipe clean with a dry cloth. Do not attempt to open the Mother E-Base™ or Daughter E-Base™. To honor the warranty, bases should only be opened and serviced by Invitrogen.
Introduction
The E-Holder™ Platform is designed to hold E-Gel® 48 and 96 gels during robotic loading. Use the E-Holder™ when you need to load multiple gels on a robotic platform while other gels are running on the E-Base™
Note: The E-Holder™ is not a power supply unit, cannot be connected to an electrical outlet, and cannot be used to run E-Gel® 96 gels.
To obtain the best results, run E-Gel® 48 or96 gels on the Mother E Base™ or Daughter E Base™ within 15 minutes after loading on E-Holder™.
Procedure
1% E-Gel® 48 Gel
Results obtained using a 1% E-Gel® 48 gel is shown in the figure below. The gel was electrophoresed for 23 minutes using the standard conditions described in this manual and imaged using a KODAK EDAS290 system.
You can use a mini transilluminator to view the bands. You may vary the amount of markers loaded on the gel to improve gel imaging.
The gel contains following samples:
| Lane | Sample |
|---|---|
| 1, 24, M (lower left, lower right) | High Mass DNA Ladder (4 µl/well) |
| 2, 3, 22, 23, 32, 33, 36, 37, 40, 41 | PCR product, 317 bp (100 ng/well) |
| 4, 5, 20, 21, 30, 31,42, 43 | PCR product, 1 kb (100 ng/well) |
| 6, 7, 18, 19, 28, 29, 44, 45 | PCR product, 3 kb (100 ng/well) |
| 8, 9, 12, 13, 16, 17, 26, 27, 46, 47 | PCR product, 9 kb (100 ng/well) |
| 10, 11, 14, 15, 34, 35, 38, 39 | E-Gel® High Range DNA Ladder (10 µl/well) |
| 25, 48, M (upper right, upper left) | 1 Kb Plus DNA Ladder (0.5 µg/well) |
2% E-Gel® 48 Gel
Results obtained using a 2% E-Gel® 48 gel is shown in the figure below. The gel was electrophoresed for 20 minutes using the standard conditions described in this manual and imaged using a KODAK EDAS290 system.
You can use a mini transilluminator to view the bands. You may vary the amount of markers loaded on the gel to improve gel imaging.
The gel contains following samples:
| Lane | Sample |
|---|---|
| 1, 24, M (lower left, lower right) | 100 bp DNA Ladder (0.4 µg/well) |
| 2, 3, 22, 23, 34, 35, 38, 39 | PCR product, 150 bp (100 ng/well) |
| 4, 5, 8, 9, 20, 21, 40, 41 | PCR product, 240 bp (100 ng/well) |
| 6, 7, 17, 18, 30, 31, 41, 42 | PCR product, 317 bp (100 ng/well) |
| 8, 9, 16, 17, 28, 29, 44, 45 | PCR product, 1 kb (100 ng/well) |
| 10, 11, 14, 15, 26, 27, 45, 47 | PCR product, 3 kb (100 ng/well) |
| 12, 13, 36, 37 | E-Gel® 96 Low Range Quantitative Ladder (10 µl/well) |
| 25, 48, M (upper left, upper right) | 50 bp DNA Ladder (0.4 µg/well) |
4% E-Gel® 48 Gel
Results obtained using a 4% E-Gel® 48 gel is shown in the figure below. The gel was electrophoresed for 20 minutes using the standard conditions described in this manual and imaged using a KODAK EDAS290 system. You can use a mini transilluminator to view the bands. You may vary the amount of markers loaded on the gel to improve gel imaging.
The gel contains following samples:
| Lane | Sample |
|---|---|
| 46, 47, M (upper left) | 10 bp DNA Ladder (1 µg/well) |
| 1, 20, 21, 38, 39, 48, M (lower right) | 25 bp DNA Ladder (0.5 µg/well) |
| 24, 25, M (lower left) | 50 bp DNA Ladder (0.5 µg/well) |
| 26, 27, M (upper right) | 100 bp DNA Ladder (0.5 µg/well) |
| 8, 13, 14, 34, 35 (10 µl/well) | E-Gel® Low Range Quantitative DNA Ladder |
| 2,3 | Synthetic 21-mer siRNA (short interfering RNA, 100 ng/well) |
| 6,7 | dsRNA diced (cut) with Dicer enzyme (100 ng/well) |
| 4,5 | Undiced dsRNA (100 ng/well) |
| 22 | ssDNA, 60 mer (200 ng/well) |
| 23 | ssDNA (lane 22) annealed to form dsDNA, 60 bp (100 ng/well) |
| 9, 10, 44, 45 | PCR product Hinf I cut |
| 11, 12, 36, 37 | PCR product Aat II cut |
| 15, 28, 29 | PCR product, 40 bp (100 ng/well) |
| 16, 30, 31 | PCR product, 72 bp (100 ng/well) |
| 17, 32, 33 | PCR product, 150 bp (100 ng/well) |
| 18, 40, 41 | PCR product, 240 bp (100 ng/well) |
| 19, 42, 43 | PCR product, 317 bp (100 ng/well) |
The table below provides some solutions to possible problems you might encounter during the electrophoresis of E-Gel® 48 and 96 agarose gels.
Problem | Cause | Solution |
No current
|
Daughter E-Base
™ used without Mother E-Base
™ |
Do not use the Daughter E-Base
™ without a Mother E-Base
™. The Daughter E-Base
™ does not have an electrical plug to connect to an electrical outlet.
|
Copper contacts in the base are damaged due to improper use
|
Make sure the copper contacts in the base are intact.
| |
Expired or defective gel cassette used
|
Use fresh gel cassette. Use properly stored gels before the specified expiration date.
| |
Gel cassette is not inserted properly into a base
|
Remove cassette and reinsert; a red light illuminates at the lower left corner of the base, a fan in the base begins to run, and digital display indicates time for a selected program or last time setting (Ready Mode) when the gel is properly inserted into the base.
| |
Poor resolution or smearing of bands
|
Sample is overloaded
|
Do not load more than 20-100 ng of sample DNA per band. Less DNA is required since E-Gel
® agarose gels are thinner.
|
High salt concentration
|
Dilute your high-salt samples as described.
| |
Very low volumes of sample loaded or sample was not loaded properly
|
Avoid introducing bubbles while loading the samples. Bubbles will cause band distortion.
Load the recommended sample volume based on the gel type and loading method.
For proper band separation, we recommend keeping sample volumes uniform. Load an equal volume of sample buffer containing the same salt concentration as your sample into the empty wells.
| |
Gel was not electrophoresed immediately after sample loading
|
For best results, run the gel within 15 minutes of sample loading.
If you cannot run the gel immediately after sample loading, use the Two-Step Loading method.
If you are using the E-Holder
™, program your robotic system to load the gel 5 minutes before the end of the previous gel’s run.
| |
A1 tip not aligned
|
Be sure to align the A1 tip properly prior to loading your samples on an E-Gel
®96 gel.
| |
Expired gel used
|
Use properly stored gels before the specified expiration date.
| |
Longer electrophoresis run time or high current during the run
|
Longer run times cause an increase in the current, resulting in poor band migration. Do not run the gel longer than the recommended time for each gel type.
| |
Uneven run on E-Gel
®48 gels
|
Differential salt concentration in adjacent lanes
|
Be sure to load 15 µl of sample buffer containing the same salt concentration as the sample into any remaining empty wells.
Keep all sample volumes uniform.
|
Slanted bands in marker lanes on E-Gel
®48 gels
|
Differential salt concentrations in adjacent lanes
|
Prepare the marker in a buffer containing the same salt concentration as the samples.
|
Sample leaking from the wells
|
Sample is overloaded
|
Be sure to load the recommended volume of sample per well.
|
Use the Two-Step Loading method.
| ||
Wells damaged during comb removal
|
Be sure to remove the comb gently without damaging the wells.
| |
Over-run the gel or need more time to run gel
|
Accidentally selected a different program
|
Select EG if you are using E-Gel
®96 gels. For E-Gel
®48 gels, select EG and then manually change the time to 20 minutes.
If you accidentally selected a different program and are at the beginning of the run, stop the run and select the desired program.
If you are well into the run, check the gel to see where the loading dye is running. Estimate the amount of time remaining and then manually stop the run.
|
Failure Mode indicated by a steady red and continuous rapid beeping, and flashing “ER” on an E-Base
™ |
Defective cassette
|
Disconnect the base and replace gel cassette with a fresh gel cassette.
Press and release the power button to return to Ready Mode.
|
Cold cassette
|
Use a room temperature cassette stored at room temperature. Avoid storing gel cassettes at 4°C.
| |
Improper operating conditions
|
Use the E-Base
™ at room temperature (20-25°C).
|
Introduction
A quick reference guide for operating the Mother E-Base™ and Daughter E-Base™ is provided below. Operating modes and electrophoresis runs are described.
Mode | Action | Sound | Light | Digital Display |
Base plugged in
|
Mother E-Base
™ connected to an electrical outlet
|
1 beep
|
No light if a cassette is not inserted, or red light if a cassette is inserted
|
Without gel cassette
-EP, last program used (EP or EG) With gel cassette in
-last time setting
|
Ready (with no current flowing through gel)
|
Gel cassette inserted into a base
|
--
|
Steady red
|
Default time setting (12 minutes for EG, 14 minutes for EP, or last time setting)
|
Run
|
Press and release the pwr/prg button
|
--
|
Steady green
|
Count down time
|
End of run
|
Automatic
|
Continuous beeping for 2 minutes followed by a single beep every minute
|
Flashing red until the time button is pressed
|
Negative time display (00 to
-19 minutes) |
Run ends after an external power failure
|
Automatic
|
Continuous beeping for 2 minutes followed by a single beep every minute
|
Alternating red and green
|
Negative time display (00 to
-19 minutes) |
Pause (manually end the run)
|
Press and release the pwr/prg button during the run
|
--
|
With gel cassette in - steady red
Without gel cassette - no light
|
Flashing time display
|
Return to Ready mode after an automatic stop
|
Press and release the pwr/prg button
|
--
|
Steady red
|
Last time setting
|
Restart after a manual stop
|
Press and release the pwr/prg button
|
--
|
Steady green
|
Count down time
|
Return to Ready mode after a manual stop
|
Press and hold the pwr/prg button
|
--
|
With gel cassette in – steady red
Without gel cassette – no light
|
With gel cassette in –last time setting
Without gel cassette - last program setting
|
Failure
|
Press and hold pwr/prg button for 2 seconds and remove gel from the base
|
Continuous loud beeping
|
Flashing “ER”
| |
No cassette
|
--
|
--
|
--
|
EP, last program used (EP or EG)
|
Time setting
|
With gel cassette in - Press and release the time button
|
--
|
With gel cassette – steady red
|
Time increases by 1 minute increments
|
With and without gel cassette - Press and hold the time button
|
--
|
With gel cassette in – steady red
Without gel cassette – no light
|
Time increases continuously and automatically stops at 00
| |
Program setting
|
Press and release the pwr/prg button when no cassette is inserted into the E-Base™ to select the desired program
|
1 beep
|
No light
|
Selected program EP or EG
|
The Mother E-Base™ and Daughter E-Base™ comply with the Underwriters Laboratories Inc. regulation and the European Community Safety requirements. Operation of the E-Gel® bases is subject to the following conditions:
Ethrog Biotechnologies Ltd., an Invitrogen company, is the manufacturer and owner of the UL file. For more information, contact:
Ethrog Biotechnologies Ltd.
Ness-Ziona Science Park
Bldg 14, P.O. Box 444
Ness-Ziona, Israel 74103![]()
The Caution symbol denotes a risk of safety hazard. Refer to accompanying documentation.![]()
Double Insulation
Class II product
E-Gel® PowerBase
The E-Gel® PowerBase™ complies with the Underwriters Laboratories Inc. regulation and is listed under file no. E189045 in the U.S. and Canada.
This device complies with part 15 of the FCC Rules. Operation is subject to the following two conditions: