Technical Resources - Media Formulations
11130 - MEM Amino Acids Solution (50X) liquid
ComponentsMolecular WeightConcentration (mg/L)mM
Amino Acids
L-Arginine hydrochloride211.06320.029.952606
L-Cystine240.01200.05.0
L-Histidine hydrochloride-H2O210.02100.010.0
L-Isoleucine131.02620.020.0
L-Leucine131.02620.020.0
L-Lysine hydrochloride183.03625.019.808743
L-Methionine149.0755.05.067114
L-Phenylalanine165.01650.010.0
L-Threonine119.02380.020.0
L-Tryptophan204.0510.02.5
L-Tyrosine181.01800.09.944752
L-Valine117.02340.020.0
Verweis
Related Products:
11120 - MEM Vitamin Solution (100X) liquid
Cat No: 11120052
11140 - MEM Non-Essential Amino Acids Solution
Cat No: 11140050 , 11140076