Search
Whether you are using a Fisher Chemical or thermo scientific product, you can trust Thermo Fisher Scientific’s range of chemicals to fulfil your research and life sciences needs. Our array of chemicals are available in varying purities and pack sizes.
Already using an existing Thermo Fisher Scientific product of instrument? For example, MagMAX kits, Kingfisher, taqpath, etc., we have complementing solvents and solutions such as the 80% Molecular Biology grade Ethanol for you. The table below outlines our top selling solvents in the biopharmaceutical and biotechnology industries.
The tables below show some of our popular products in the biopharmaceuticals segment. Find out more from us today to place an order.
Catalog Number | Description |
BP24384
| 1 × PBS 4L |
BP3991
| 10 × Phosphate buffered saline (PBS) 1 L |
BP39920
| 10 × Phosphate buffered saline (PBS) 20 L |
BP3994
| 10 × Phosphate buffered saline (PBS) 4 L |
BP24711
| 10 × TBS Buffer pH7.4 1 L |
BP6651
| 10 x PBS powder concentrate, makes 2 x 1L |
BP82011
| 70% Ethanol 1 L |
T08204K7
| 80% Ethanol 4 L |
BP2818100
| Ethanol, Absolute (200 Proof) 100 mL |
BP28184
| Ethanol, Absolute (200 Proof) 4 L |
BP2818500
| Ethanol, Absolute (200 Proof) 500 mL |
BP11704
| Acetonitrile anhydrous (DNA synthesis) 4 L |
BP1423500
| Agar granulated 500 g |
BP160500
| Agarose Low EEO 500 g |
BP1356500
| Agarose MB prep 500 g |
BP1160500
| DMF, Dimethylformamide, Sequencing 500mL |
BP231100
| DMSO Dimethyl sulfoxide 100 mL |
BP17225
| Dithiothreitol, DTT 25 g |
Catalog Number | Description |
BP2291
| Glycerol 1 L |
BP2294
| Glycerol 4 L |
BP3815
| Glycine 5 kg |
BP1781
| Guanidine hydrochloride 1 kg |
BP3101
| HEPES, molecular biology 1 kg |
BP310100
| HEPES, molecular biology 100 g |
BP26184
| Isopropanol, molecular biology 4 L |
BP1752I400
| Phenol / chloroform / isoamyl alcohol 25:24:1 mixture pH 6.7/8.0 400 mL |
BP358212
| Sodium chloride 2.5 kg |
BP166500
| Sodium dodecyl sulfate (SDS) 500 g |
BP1521
| Tris base 1 kg |
BP1525
| Tris base 5 kg |
BP337500
| Tween 20 500 mL |
BP169500
| Urea, molecular biology 500 g |
BP5611
| Water (RNA grade) 1 L |
BP28191
| Water, molecular biology grade 1 L |
BP1422500
| Yeast Extract Powder, 500 g |
Download our Life Sciences related brochures today!
As chemical needs differ from customer to customer, one of the following situations in technical requirements may occur to you:
1. Blend of chemicals
2. Specified labeling
3. Bulk orders
4. All other chemicals not listed on our brochure
You can have confidence in our custom & bulk synthesis team to supply what you need. Chat with us today to find out more about how we can become your one-stop solution for all your semiconductor applications.
Fisher Bioreagents products deliver an end-to-end solution that can meet your most demanding electrophoresis requirements. You can depend on our expertise in electrophoresis instruments along with ultrapure reagents that are pre-qualified for your applications.
Sodium dodecyl sulfate – polyacrylamide gel electrophoresis (SDS-PAGE) is the most direct method for assessing in a fast and reproducible manner, the relative molecular weight (Mr) of denatured polypeptide chains and the purity of a protein preparation. In SDS-PAGE, the sample to be applied to the gel is first treated with the anionic detergent SDS which denatures the proteins in the sample and binds tightly to the protein molecules. The SDS molecules confer a relatively uniform negative charge to the polypeptide in proportion to its length. When an electric current is applied across the gel, all proteins will migrate through the gel matrix toward the anode. In this way, SDS-PAGE separates proteins according to size because the SDS-coated proteins have a uniform charge:mass ratio. Proteins with less mass travel more quickly through the gel than those with larger mass because of the sieving effect of the gel matrix.
Preparation of Polyacrylamide Stacking and Separating Gels (SDS-PAGE)
Separating Gel (Total Volume, 15mL)1
Final % Acrylamide in Gel2 Stock Solutions3 |
5
|
6
|
7
|
7.5
|
8
|
9
|
10
|
12
|
13
|
15
|
30% Acrylamide/0.8% Bis-Acrylamide |
2.50mL
|
3.00mL
|
3.50mL
|
3.75mL
|
4.00mL
|
4.50mL
|
5.00mL
|
6.00mL
|
6.50mL
|
7.50mL
|
4X Tris•Cl, pH 8.8 |
3.75
|
3.75
|
3.75
|
3.75
|
3.75
|
3.75
|
3.75
|
3.75
|
3.75
|
3.75
|
H2O4 |
8.60
|
8.10
|
7.60
|
7.35
|
7.10
|
6.60
|
6.10
|
5.10
|
4.60
|
3.60
|
10% SDS |
0.15
|
0.15
|
0.15
|
0.15
|
0.15
|
0.15
|
0.15
|
0.15
|
0.15
|
0.15
|
10% Ammonium Persulfate5 |
0.05
|
0.05
|
0.05
|
0.05
|
0.05
|
0.05
|
0.05
|
0.05
|
0.05
|
0.05
|
TEMED |
0.01
|
0.01
|
0.01
|
0.01
|
0.01
|
0.01
|
0.01
|
0.01
|
0.01
|
0.01
|
Procedure for Gel Preparation
In a 25mL sidearm flask, mix the given volumes of Acrylamide/Bis-Acrylamide solution, Tris•HCl buffer, and H2O. Degas under vacuum 10 to 15 minutes. Add the SDS solution, Ammonium Persulfate solution, and TEMED. Swirl gently to mix. Use immediately.
1. These volumes are adequate for a gel of dimensions 0.75cm x 14cm x 14cm. (The recipes are based on the SDS (denaturing)-continuous buffer system of Laemmli (1970).)
2. The % acrylamide selected for the separating gel will depend on the molecular sizes of the proteins being separated.
3. Recipes for the stock solutions appear earlier in this section.
4. All reagents and solutions used in this protocol must be prepared with distilled deionized water.
5. Store at 4°C (maximum 5 days).
Product code | Product description / Concentration | Pack size |
TG, Tris-Glycine | ||
BP1306
|
Tris-Glycine, 10X Solution
|
1 L, 4 L
|
TGS, Tris-Glycine-SDS | ||
BP1341
|
Tris-Glycine-SDS, 10X Solution
|
1 L, 4 L
|
BP1398
|
Tris-Glycine-SDS, 5X Powder*
|
92 g (1L equivalent)
|
SDS, Sodium Dodecyl Sulfate | ||
BP2436
|
10% Sodium Dodecyl Sulfate Solution
|
200 mL, 1 L
|
BP1311
|
20% Sodium Dodecyl Sulfate Solution
|
200 mL, 1 L
|
PBS, Phosphate Buffered Saline | ||
BP399
|
PBS, Phosphate Buffered Saline, 10X Solution
|
500 mL, 1 L, 4 L, 20 L
|
TBS, Tris-Buffered Saline | ||
| BP2471 | TBS, Tris Buffered Saline, 10X Solution, pH 7.4 | 100 mL, 500 mL, 1 L |
| *Pre-weighted powder to make 1L of 5X buffer | ||
Product code | Product description | Pack size |
BP170
| Acrylamide (White, Needle-like Crystals/Electrophoresis) |
100 g, 500 g, 5 kg
|
BP1364
| Acrylamide: Bis-Acrylamide, dry powder mix, 19:1 (5% cross-linker) |
100 g
|
BP1406
| Acrylamide: Bis-Acrylamide, 40% solution, 19:1 (5% cross-linker) |
1 L
|
BP1408
| Acrylamide: Bis-Acrylamide, 40% solution, 29:1 (3.3% cross-linker) |
1 L
|
BP1410
| Acrylamide: Bis-Acrylamide, 40% solution, 37.5:1 (2.6% cross-linker) |
1 L
|
BP179
| Ammonium Persulfate (Colorless-to-White Crystals/Electrophoresis) |
25 g, 100 g
|
BP150
| TEMED (Electrophoresis) |
20 g, 100 g
|
Product code | Product description | Pack size |
BP571
| CHAPS (White Crystalline Powder) |
1g, 5g
|
BP166
| Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis |
100 g, 500 g, 5 kg
|
BP2436
| 10% Sodium Dodecyl Sulfate Solution |
200 mL, 1 L
|
BP1311
| 20% Sodium Dodecyl Sulfate Solution |
200 mL, 1 L
|
BP151
| Triton™ X-100 (Electrophoresis) |
100 mL, 500 mL
|
BP337
| Tween 20 |
100 mL, 500 mL
|
BP338
| Tween 80 |
500 mL
|
BP585
| N-Ocytl-B-D-Glucopyranoside |
1 g, 5 g, 25 g
|
Agarose is a linear polysaccharide composed of alternating residues of D- and L-galactose joined by glycosidic linkages. Agarose forms gels that are both porous and resilient. These gel properties provide a sieving matrix that allows the electrophoretic separation of charged macromolecules such as DNA or RNA according to size. Compared to polyacrylamide gel, agarose has a lower resolution but wider range of separation. Lower grades of agarose can be contaminated with other polysaccharides, salts, and proteins. Such impurities can alter the gelling/melting temperature of agarose solutions or affect the ability to use the recovered nucleic acid sample in a post-electrophoresis application.
All Fisher BioReagents® agarose are DNase and RNase-free to ensure optimal results for your nucleic acid application. Fisher BioReagents offers three different grades of agarose that are functionally tested and pre-qualified for specific applications.
Agarose grades used in electrophoresis of nucleic acids:
1. Genetic Analysis Grade—agarose that yields biologically active DNA or RNA. Testing includes enzymatic performance measurements.
2. Molecular Biology Grade—suitable for analytical separation of DNA or RNA.
3. PCR Grade—the original agarose for analytical separation of PCR amplicons (<1kb).
1. The size of DNA or RNA fragments to be analyzed (see graph below).
Cat. No.
| Agarose Separation Ranges |
500bp to 23 kb
| |||||||||||||||
BP160
| Low EEO/Multipurpose |
200bp to 25kb
| |||||||||||||||
BP165
| Low Melting/Nucleic Acid Recovery |
500bp to 25kb
| |||||||||||||||
BP1365
| Broad Separation Range for DNA/RNA |
50bp to 1kb
| |||||||||||||||
BP1360
| Low Melting <1kb DNA/RNA |
50bp to 1kb
| |||||||||||||||
BP2410
| Intermediate Melting |
0
|
100
|
200
|
300
|
400
|
500
|
600
|
700
|
800
|
900
|
1000
|
1100
|
1200
|
23K
|
25K
| |
2. The type of downstream application that will follow electrophoretic separation (e.g., cloning procedures directly from re-melted agarose or in-gel reaction).
Type of Agarose | Low EEO | Low Melting >200bp | Low Melting >1000bp | Wide Separation Range | PCR Grade |
Cat No. | BP160 | BP165 | BP1360 | BP1356 | BP2410 |
Recovery of DNA and RNA
|
X
|
X
|
X
|
X
|
X
|
Southern and Northern Blots
|
X
| ||||
DNA/RNA separation 50bp to 1kb
|
X
|
X
| |||
DNA/RNA separation >1kb
|
X
|
X
| |||
PCR fragment analysis
|
X
|
X
|
X
|
X
|
X
|
In-gel reactions (ligation, transformations, PCR)
|
X
| ||||
Colony lifts
|
X
| ||||
Agarose grade
|
Molecular Biology
|
Molecular Biology
|
Genetic Analysis
|
Genetic Analysis
|
PCR
|
Available pack sizes
|
100 g, 500 g
|
25 g
|
100 g
|
100 g, 500 g
|
100 g
|
Two buffers commonly used for DNA agarose electrophoresis are Tris-acetate with EDTA (TAE; 40mM Tris-acetate, 1mM EDTA) and Tris-borate with EDTA (TBE, 89mM Tris-borate, 2mM EDTA). Because the pH of these buffers is neutral, the phosphate backbone of DNA has a net negative charge and migrates toward the anode. TAE and TBE have different properties which makes one more suitable than the other for a specific purpose.
MOPS is a commonly used buffer system for RNA electrophoresis using formaldehyde or formamide denatured RNA. It is important to use RNase-free chemicals, water and containers when preparing the buffer solution. The typical formulation of a 10X MOPS running buffer is 0.4M MOPS (pH 7.0), 0.1M sodium acetate, and 0.01M EDTA. The denaturing system chosen depends on the purpose of the RNA experiment and the size of the RNA fragment being separated. Formaldehyde denaturation is suitable if RNA samples are to be recovered. Formamide denaturation is suitable if the RNA needs to retain its biological activity.
Buffer | Suggested Uses | Properties |
TAE
|
DNA recovery.
Electrophoresis of large DNA (>12kb). |
Low buffering capacity.
Recirculation may be necessary for extended run times (>6 hr.) |
TBE
|
Electrophoresis of small DNA (<1kb).
Increased resolution of small DNA (<1kb). |
Decreased DNA mobility.
High buffering capacity – no recirculation required for extended run times. |
MOPS
|
Electrophoresis of formaldehyde denatured RNA.
|
Buffer is low in ionic strength.
Recirculation of buffer may be necessary. |
Suggested Agarose Concentrations: The optimal gel concentration depends on the size of the DNA fragments to be resolved.
Cat No. | Main Application | DNA Size Range in Base Pairs | Final Agarose Concentration % w/v 1x TAE Buffer | Final Agarose Concentration % w/v 1x TBE Buffer |
BP1360
|
Low melting temperature agarose. Certified recovery of small nucleic acid fragments. Outstanding resolution.
|
500 – 1,000
|
2.5
|
2.0
|
150 – 700
|
3.0
|
2.5
| ||
100 – 450
|
3.5
|
3.0
| ||
70 – 300
|
4.0
|
3.5
| ||
10 – 100
|
4.5
|
4.0
| ||
8 - 50
|
5.0
|
4.5
| ||
BP165
|
Low melting temperature agarose. Broad separation range. Ideal for DNA and RNA recovery after electrophoretic separation.
|
500 – 25,000
|
0.75
|
0.70
|
300 – 20,000
|
1.00
|
0.85
| ||
200 – 12,000
|
1.25
|
1.00
| ||
150 – 6,000
|
1.50
|
1.25
| ||
100 – 3,000
|
1.75
|
1.50
| ||
50 – 2,000
|
2.00
|
1.75
| ||
BP1356 BP160
|
Suitable for routine nucleic acid electrophoresis applications with broad separation range.
|
1,000 – 23,000
|
0.60
|
0.50
|
800 – 10,000
|
0.80
|
0.70
| ||
400 – 8,000
|
1.00
|
0.85
| ||
300 – 7,000
|
1.20
|
1.00
| ||
200 – 4,000
|
1.50
|
1.25
| ||
100 – 3,000
|
2.00
|
1.75
|
Product code | Product description | Pack size |
TBE Tris-Borate-EDTA | ||
BP2430
| TBE Buffer, Tris-Borate-EDTA, 1X Solution, Electrophoresis |
1 L, 4 L, 20 L
|
BP1333
| TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis |
1 L, 4 L, 20 L
|
TAE, Tris-Acetate-EDTA | ||
BP1335
| TAE Buffer, Tris-Acetate-EDTA, 10X Solution, Electrophoresis |
500 mL, 1 L, 4 L, 20 L
|
BP1332
| TAE Buffer, Tris-Acetate-EDTA, 50X Solution, Electrophoresis |
500 mL, 1 L, 4 L, 20 L
|
MOPS | ||
BP308
| MOPS (Fine White Crystals/Molecular Biology) |
100 g, 500 g
|
Formaldehyde | ||
BP531
| Formaldehyde (37% by Weight/Molecular Biology) |
25 mL, 500 mL
|
Tris Base | ||
BP152
| Tris Base (White Crystals or Crystalline Powder/Molecular Biology) |
500 g, 1 kg, 5 kg, 10 kg, 25 kg
|
Glacial Acetic Acid | ||
BP2401
| Acetic Acid, Glacial (Certified ACS Plus) |
500 mL, 2.5L
|
| Boric Acid | ||
BP168
| Boric Acid (Crystalline/Electrophoresis) |
500 g, 1 kg
|
EDTA | ||
BP120
| EDTA, Disodium Salt Dihydrate (Crystalline Powder/Electrophoresis), |
500 g, 1 kg
|
Liquid chromatography-Mass Spectroscopy (LC-MS) can be used to characterize virus proteins and the recombinant proteins produced by viral vectors that both are essential throughout the vaccine development and many manufacturing workflows. It is also used for Omics-based diagnostic tests of virus protein/peptide biomarkers complement the RT-PCR based methods.
Here are some of our solvents and reagents used to support these LC-MS workflow applications:
Application | Product Description | Cat No. | Pack Size |
Protein Precipitation & Digestion | |||
Acetone
|
Acetone, Optima™, for HPLC and GC
|
A929
|
1 L, 4 L
|
Trifluoroacetic acid (TFA)
|
Trifluoroacetic Acid, Optima™ LC/MS Grade
|
A116
|
0.5 mL, 1 mL, 2 mL, 50 mL
|
Formic Acid
|
Formic Acid, 99.0+%, Optima™ LC/MS Grade
|
A117
|
0.5 mL, 1 mL, 2 mL, 50 mL
|
LC-MS/MS Mobile Phase | |||
0.1% FA in Water
|
Water with 0.1% Formic Acid (v/v), Optima™
|
LS118
|
500 mL, 1 L, 2.5 L, 4 L
|
0.1% FA in Acetonitrile
|
Acetonitrile with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade
|
LS120
|
500 mL, 1 L, 2.5 L, 4 L
|
2-propanol
|
Isopropanol, Optima™ LC/MS Grade
|
A461
|
500 mL, 1 L, 2.5 L, 4 L
|
Acetonitrile
|
Acetonitrile, Optima™ LC/MS Grade
|
A955
|
500 mL, 1 L, 2.5 L, 4 L
|
Methanol
|
Methanol, Optima™ LC/MS Grade
|
A456
|
1 L, 2.5 L, 4 L
|
Water
|
Water, Optima™ LC/MS Grade
|
W64
|
500 mL, 1 L, 2.5 L, 4 L
|
Buffer for stabilization of whole protein units | |||
Ammonium acetate
|
Ammonium acetate, Optima™ LC/ MS grade
|
A114
|
50 g
|
To view our extensive catalog on Liquid Chromatography solvents, head on over to our Analytical Solvents page.
For Research Use Only. Not for use in diagnostic or therapeutic procedures.
For Research Use Only. Not for use in diagnostic or therapeutic procedures.