What is a combinatorial DNA library?
A combinatorial DNA library is a collection of DNA sequences where multiple nucleotide positions are mutated within the sequence, allowing for various combinations of mutations at different positions. This process enables the exploration of a wide range of genetic variations. Based on true rational design, combinatorial DNA libraries can achieve a maximum diversity to screen for and identify synergistic beneficial mutations, such as affinity maturation via phage display libraries.
Applications of combinatorial DNA library
Combinatorial DNA libraries are utilized in various fields such as drug discovery, protein engineering, and gene function studies, enabling researchers to explore genetic variations and identify optimal sequences for various applications, including:
  • Optimizing antibody affinity: Facilitate antibody affinity optimization through affinity maturation
  • Humanizing antibodies: Modify antibodies to be more human-like to reduce immune responses
  • Modifying antibody specificity: Alter the specificity of antibodies to target different antigens
  • Improving protein properties: Help enhance physiochemical properties such as solubility, heat stability, and activity in industrial environments
  • Enhancing function: Help improve promoter activity or enzymatic properties
  • Prolonging therapeutic half-life: Help extend the half-life of therapeutics for in vivo applications
  • Improving enzyme function: Help optimize enzyme function for industrial use
  • Modifying enantioselectivity: Change the enantioselectivity of enzymes
  • Altering patented sequences: Induce changes to modify proteins from patented sequences
Why choose GeneArt combinatorial DNA libraries?
All GeneArt Combinatorial Library products are sequenced and statistically analyzed to help ensure that they meet the following quality benchmarks:
  • Sequencing of up to 96 individual transformants to verify that customer’s specifications regarding amino acid content and distribution have been met, and to verify sequence integrity of unmutated portions of the construct
  • Bulk sequencing to verify that nondegenerate portions of the construct have the correct sequence and randomized portions meet customer’s specifications
  • Real-time PCR prior to amplification to verify that library diversity objectives have been met
  • Optional next-generation sequencing quality control (additional fee applies)
Quality control:
Sequencing of backbone region (100% correctness)
Quality control:
RT-PCR
Quality control:
Bulk sequencing
Quality control:
Peer group sequencing, NGS (optional)
Quality control:
Sequencing of backbone region (100% correctness)
Quality control:
Bulk sequencing
Quality control:
Peer group sequencing, NGS (optional)
GeneArt combinatorial libraries: Selection guide
 GeneArt Strings DNA librariesGeneArt combinatorial librariesGeneArt combinatorial libraries with next-generation sequencing
Advantage
  • Cost efficient
  • Fast service
  • Flexible design
  • Better control over mutations
  • Flexible design
  • Better control over mutations
  • Enhanced QC detection of low abundance codons
Design flexibility+
Full IUPAC code available; however limited control over occurring amino acids
+++
TRIM technology allows for the accurate determination of occurring amino acids and ratios
+++
TRIM technology allows for the accurate determination of occurring amino acids and ratios
Correctness+
Gene synthesis process used, more unintended mutations occur
+++
Error free template enables best possible correctness of results
+++
Error free template enables superior correctness of results
Complexity<1011<1011
<1012 optional
<1011
<1012 optional
Cost$$$$$$
Production time+++
10–15 business days
+
4–6 weeks
+
4–6 weeks
 Learn moreSee aboveSee above
Delivery formats
GeneArt ready-to-clone combinatorial library
  • 2–5 µg of linear DNA ready to clone via 5' and 3' restriction sites
  • Non-amplified library, 17–170 fmol (1010–1011 specimen variant diversity) depending on library diversity
  • Amplification primers
GeneArt cloned combinatorial library
  • >30 µg of plasmid DNA cloned into the vector of your choice
  • 109 transformation rate
  • 12 aliquots of 0.5 mL glycerol stocks
  • Non-amplified library, 17–170 fmol (1010–1011 specimen variant diversity) depending on library diversity
  • Amplification primers
Get started with your DNA combinatorial library project
Please download the GeneArt Combinatorial Libraries Custom Service Requirements Form to submit project information. For secure data transfer, please register at our customer portal.
Libraries made with conventional technology by the incorporation of degenerate codons (NNS, VNS, etc.) are also available. Please use the above-mentioned questionnaire for both technologies.
For further information regarding this service or the ordering process, please contact geneartsupport@thermofisher.com.
Optimize your combinatorial DNA library with TRIM technology
Besides randomization via degenerate codons (e.g., NNS), the sequences of GeneArt Combinatorial DNA Libraries can also be diversified using preassembled trinucleotide building blocks (trinucleotide mutagenesis (TRIM) technology) within the chemical synthesis process, to achieve significantly higher quality than by using conventional technologies. This allows for complete customization of the amino acid composition at randomized sites and thus avoids the occurrence of unwanted stop codons or amino acids. Achieve an extra level of QC by requesting optional next-generation sequencing of your combinatorial DNA libraries using the Thermo Scientific Ion Torrent sequencing platform (see case study below).
Benefits of using TRIM technology
  • Rational diversity: Control important features of the library using TRIM technology to create rational diversity where it is likely to have the most impact
  • Screening efficiency: Helps significantly reduce screening efforts compared to conventional mutagenesis methods, thus helping to save time and budget
  • Success rate: Customized design allows for the inclusion of only substitutions known to contribute to a certain property, which increases the likelihood of screening success
  • Flexibility: Even the ratio of allowed amino acids can be accurately controlled, for example, with ratios reflecting exactly the ones found in humans
  • Accuracy: TRIM significantly reduces frameshift mutations and "abolishes" the occurrence of stop codons which makes the screening of libraries more efficient. Ancillary mutation rates achieved are typically in a correctness range of >82%
  • Achievable library diversity: Combinatorial DNA libraries can achieve diversities of up to 1012
  • Speed: Our capacities allow for fast production times that are project-specific
Schematic illustrating sequence randomization using degenerate codons (NNS) versus TRIM technology
Figure 1. Comparison of sequence randomization using degenerate codons (e.g. NNS, N: all 4 bases at 25%; S: G & C at 50% each) vs. TRIM technology.
Combinatorial DNA libraries with NGS quality control—a case study
Highlights:
  • Accurate: Determination of library correctness, amino acid distribution, and uniqueness of clones
  • Comprehensive: Delivers millions of reads for analysis
  • Reliable: Sequence at clonal level with a proprietary technology and software solution
A GeneArt Combinatorial DNA Library from our production workflow was characterized using our current standard quality control protocol (“CE-sequenced”) and our novel next-generation sequencing (NGS)-based quality control for combinatorial DNA libraries (“NGS-sequenced”). The expected amino acid distribution is shown in the right-most panel (“expected”); in this case cysteines were not desired in the randomized positions. Our current standard QC protocol entails CE sequencing of at least 96 library clones and subsequent statistical analysis of the amino acid distribution at randomized positions. In this case, 304 variant clones were analyzed. In contrast, our novel NGS-based quality control for combinatorial DNA libraries on the Ion PGM (Personal Genome Machine) Sequencer allows for the analysis of tens of thousands of library clones. In this case, 41,625 clones were analyzed.
The table shows the percentage at which each amino acid appears in that position for 4 codons (X1 – X4). Note that in the CE-sequenced peer group the codons for two amino acids were not found, although they are present in the library, as shown by NGS sequencing (red and green circles).
The research leading to results regarding NGS quality control has received funding from the European Union Seventh Framework Programme (FP7/2007-2013) under grant agreement number 613931.
Table 1. Comparison of standard to NGS sequencing quality control for GeneArt Combinatorial DNA Libraries.
 Comparison of standard to NGS sequencing quality control for GeneArt Combinatorial DNA Libraries
Resources
Watch how-to videos and explainers on GeneArt Instant Designer.
Contact us
Request more information
North America and Asia: 1-800-955-6288
Ask for "GeneArt" when prompted
Europe: +49 (0)941 942 76-100
Order support:
Geneartsupport@thermofisher.com
Stay up to date on gene synthesis and protein synthesis service offerings.
Read white papers on de novo gene synthesis technologies.