Search
Thermo Scientific Talos (Scanning) Transmission Electron Microscopes are available in a variety of configurations to best suit your needs. Whether studying nanomaterials, biological samples, or analyzing semiconductor devices, having a specialized (S)TEM instrument helps ensure optimal performance and insightful data collection with high throughput for your applications. Each model offers unique features and capabilities tailored to different needs, empowering you to achieve your goals with precision and efficiency. Use the selection guide to find the right Talos (S)TEM for your applications.
For materials science applications, Talos (S)TEMs enable comprehensive characterization of materials down to the atomic scale, allowing you to elucidate the microstructure and other important properties of materials. Featuring high-resolution imaging, comprehensive elemental analysis, and in situ capabilities, the Talos TEMs empower materials scientists to unlock new insights and drive innovation in diverse fields ranging from understanding crystal defects to exploring interfaces, phase transformations, and more.
The Thermo Scientific Talos F200X Scanning Transmission Electron Microscope ((S)TEM) combines outstanding high-resolution STEM imaging with high-performance EDS. Featuring four in-column SDD Super-X EDS Detectors, it delivers 2D and 3D chemical characterization plus compositional mapping with unique cleanliness. Integrated with Thermo Scientific Velox Software, this platform generates high-accuracy and repeatable results, offering the multi-purpose versatility and heavy-duty flexibility that customers need for a diverse range of research and industrial applications.
Key features
The Thermo Scientific Talos F200S Scanning Transmission Electron Microscope ((S)TEM) provides essential high-resolution STEM imaging with features designed to enhance throughput, precision, and ease of use. As a practical alternative to the Talos F200X (S)TEM, it offers a more cost-effective solution for a wide range of applications in academic, government, and industrial research, delivering reliable performance for core imaging and analysis needs.
Key features
The Thermo Scientific Talos F200i Scanning Transmission Electron Microscope was uniquely designed for performance and productivity across a wide range of materials science applications. Featuring a more compact enclosure and a standard X-TWIN pole piece gap that enhances flexibility, it opens more opportunities for high-resolution 2D and 3D characterization, in situ dynamic observations, and diffraction applications.
Key features
Talos TEMs offer flexible solutions for academic, pathology, and pharmaceutical imaging facilities. From ultrastructural imaging of thin-sectioned cells and tissues, to nanoparticle characterization for drug development, to cryo-electron microscopy applications such as native-state imaging and single particle analysis, Thermo Scientific TEMs support your current needs and can be upgraded as your applications expand over time.
Specifically designed for semiconductor applications, the Thermo Scientific Talos F200E (S)TEM delivers fast, precise, quantitative characterization of semiconductor devices in multiple dimensions. With innovative features designed to optimize productivity, precision, and ease-of-use, the Talos F200E (S)TEM is the flexible solution for semiconductor analytical applications.
Key features
| Materials Science | Semiconductors | Life Sciences | |||||||
| Features | Talos F200x | Talos F200i | Talos F200S | Talos F200E | Talos F200C | Talos 12 | |||
| Pole piece | X-TWIN | X-TWIN | X-TWIN | C-TWIN | |||||
| Gun type | X-FEG | X-CFEG | S-FEG | X-FEG | X-CFEG | S-FEG | X-FEG | X-FEG | LaB6/W |
Thermo Fisher Scientific offers a wide range of accessories and upgrades for scanning transmission electron microscopes to provide the latest capabilities for your instrument, helping you stay at the forefront of scientific advancements in materials science research.
Optional upgrades and enhancements can keep your system state-of-the-art and help you use your instrument in the most effective and productive way possible.
| I have a | Talos G1/G2 (S)TEM | Talos G2.1 (S)TEM | Upgrade with |
| 4DSTEM/Virtual Detectors |
Win10 & NG STEM | Option | 4DSTEM on Ceta Camera |
| System Connect | t.b.f. | t.b.f. | System Connect |
| NanoMegas Ceta & HAADF | Win10 | Option | NanoMegas |
| AutoScript Software for TEM | Win10 | Option | AutoScript Software for TEM |
| Falcon 4i Detector | Win 10 | Option | Falcon 4i Detector |
| Velox Software V3.x | Win10 | Included with Win10* | |
| Smart Cam | Win10 | Included with Velox* | |
| ClickTilt | Win10 | Included with Velox* |
(*) Free with Win10
Explanation of table:
Win10 (3.17): Upgrade to Windows 10 is required for this option, last available version is 3.17 (3.18 and later not supported)
Win 10: Upgrade to Windows 10 is required for this option, no restrictions (yet) on last version (3.18 and later supported)
NG STEM: An upgrade with NG STEM (Panther STEM Detector) is required for this option
Option: An upgrade to have this option is possible without software upgrade (an upgrade of the server software may be required)
For Research Use Only. Not for use in diagnostic procedures.