Search
We offer a number of reporter systems for cell-based assays. GeneBLAzer reporter assays use a β-lactamase gene combined with a FRET-enabled substrate to provide reliable and sensitive detection in cells. CellLight products using the brightest most stable fluorescent protein colors in Baculovirus-mediated BacMam delivery system put our content or yours into cells with ease. TC-FlAsh and TC-ReAsh reagents are the smallest commercially available expression tags, offering two colors and convenience of mammalian Gateway cloning tools for live-cell work.
We offer a wide selection of fluorescence-based indicators for calcium, including Fura-2, Indo-1, Fluo-3, Fluo-4, and Rhod-3.
We offer GeneBLAzer reporter assays which consists of a β-lactamase gene combined with a FRET-enabled substrate to provide reliable and sensitive detection in cells. We also have fluorogenic substrates for β-lacatamase enzyme activity.
We offer novel technologies for cell-based fluorescent protein (FP) assays. These include organelle-specific assays (CellLight constructs), a wide variety of vectors (Gateway, TOPO and Vivid Colors reagents and kits) as well as fluorescent counterstaining reagents and kits.
Learn more about CellLight fluorescent protein labeling
The TC-FlAsH/ReAsH expression tag-based fluorescence labeling technology was developed for live-cell imaging. It is ideal for protein localization or real-time protein production studies.
For Research Use Only. Not for use in diagnostic procedures.