Search
We’ve expanded our portolio to include the Advanced Bioprocessing feeds and supplements product line. Select from a variety of feeds and supplements that are designed to enhance your cell culture up to a 2- to 5-fold increase over batch culture. Take control over traditional solubility limitations impacting total bioreactor working volume with Gibco EfficientFeed+ and GlycanTune+ Feeds. Now you can deliver superconcentrated nutrients in our Gibco Advanced Granulation Technology (AGT) dry-format for simple, rapid preparation that is designed to reduce risk.
Our Feed Kits are the perfect way to try our feeds and supplements. All kits include a user manual that provides supplementation strategies to get you started.
OneFeed improves cell performance across media and cell lines and can be used as a feed supplement to enhance growth and production of a variety of CHO lines. This flexible feed can also be added to existing processes.
Resurge formulations CD1 through CD5 compliment a variety of base media and were developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. In order to increase the versatility of the formulations, glucose and L-glutamine are not included in the final formulations. Therefore, these two components should be supplemented by users and monitored as appropriate.
Gibco Starter Paks provide samples of our most widely used peptones tailored for specific applications, including monoclonal antibodies, recombinant proteins, and vaccines, for use in both mammalian cell culture and microbial fermentation. With the 100 g samples conveniently packaged in a box, you can identify the correct supplements for your specific cell lines faster.
Follow the flow paths to help decide which feeds and supplements you should try first. To further optimize or enhance your cell culture, contact our Gibco BioProduction Services team for consultation.
![]() | ![]() | |
Gibco Dynamis Medium provides the power to achieve higher titers, faster process development, and seamless scale-up.
| Applied Biosystems GlycanAssure Glycan Analysis and Quantitation System The first glycan analysis platform that combines both high throughput and high data quality, and offers an integrated glycan analysis platform that helps save labor, time, and cost of analysis. | Feed development and optimization services. |
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing.
Explore our resources
Download Bioprocessing Product Reference Guide
For Research Use Only. Not for use in diagnostic procedures.