    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • ELISA Kits ›
          • Midkine ELISA Kits
          This product was previously sold as SKU 900-K190
          Invitrogen
          Human MDK ELISA Development Kit (ABTS), PeproTech®
          4 References
          View all (2) Midkine ELISA Kits
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          Human MDK ELISA Development Kit (ABTS)
          Group 53 Created with Sketch.
          Human MDK ELISA Development Kit (ABTS)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          Human MDK ELISA Development Kit (ABTS)
          Human Midkine ABTS ELISA standard curve (Product # 900-K190K). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Human MDK ELISA Development Kit (ABTS)
          Human MDK ELISA Development Kit (ABTS) (900-K190K)
          videoThumbnail
          ►
          Product Specifications
          Analytical sensitivity
          Lot-specific
          Assay range
          Lot-specific
          Hands-on time
          1 hr 45 min
          Time-to-result
          24 hr 30 min
          Homogenous (no wash)
          No
          Instrument
          Absorbance Microplate Reader
          Product size
          10 x 96 Tests
          Contents
          This Human Midkine ELISA Kit includes Capture Antibody, Detection Antibody, Protein Standard, and HRP-conjugate. The HRP-conjugate is packaged separately.
          Storage
          Refer to product documentation for component specific storage temperature.
          Protein name
          Midkine
          Protein family
          Growth Factors & Receptors
          Species (tested)
          Human
          Detector antibody conjugate
          Biotin
          Label or dye
          HRP
          About This Kit
          900-K190K consists of: 900-K190 (capture antibody, detection antibody, and protein standard), and AVIDIN-60UL (avidin HRP-conjugate).
          Human Midkine ABTS ELISA Development Kit contains the key components required for the quantitative measurement of natural and/or recombinant human Midkine in a sandwich ELISA format. Human Midkine ABTS ELISA Development Kit contains enough material to assay the target cytokine in at least 1,000 ELISA plate wells.
          The ABTS ELISA Buffer Kit (Product # 900-K00), sold separately, is recommended to be used in conjunction with this ABTS ELISA Development Kit for optimal performance. This buffer kit contains all the necessary components to assay ten 96-well ELISA plates (included) and contains detailed handling instructions.
          Target information
          Midkine is a retinoic acid-responsive, heparin-binding growth factor expressed in various cell types during embryogenesis. It promotes angiogenesis, cell growth, and cell migration. Midkine is also expressed in several carcinomas, suggesting that it may play a role in tumorigenesis, perhaps through its effects on angiogenesis.
          Bioinformatics
          Gene aliases : ARAP, MK, NEGF2
          Gene ID : (Human) 4192
          Gene symbol : MDK
          Protein Aliases : amphiregulin-associated protein, FLJ27379, midgestation and kidney protein, neurite growth-promoting factor 2, neurite outgrowth-promoting factor 2, retinoic acid inducible factor
          UniProt ID (Human) P21741
          Recommended Products
          IL-6 Human Matched Antibody Pair
          Because you are interested in IL-6 Human ELISA Kit.
          SIZE
          10 x 96 tests
          PRICE
          USD 400.00
          Cat # CHC1263
          guarantee_icon
          Performance Guarantee
          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0