    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
          • Order Status
          • 快速訂購
          • 登入
            登入
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • ELISA Kits ›
          • CX3CL1 ELISA Kits
          Invitrogen
          Human Fractalkine ProQuantum Immunoassay Kit
          View all (5) CX3CL1 ELISA Kits
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Additional Information:
          {{banner.description}}
          Human Fractalkine ProQuantum Immunoassay Kit
          Group 53 Created with Sketch.
          Human Fractalkine ProQuantum Immunoassay Kit
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          Human Fractalkine ProQuantum Immunoassay Kit
          How ProQuantum immunoasssays work
          (A) Antibody-antigen binding: Antibodies bind to two separate epitopes on the antigen (during a 1-hr incubation), which brings the two conjugated oligonucleotides into close proximity. (B) Ligation and amplification of signal (in a qPCR instrument): DNA ligase and a third splint oligonucleotide connector are added to ligate the two ends of the conjugated oligonucleotides, creating a 100-base DNA template. Once the ligase is inactivated at 95°C, the sample is amplified through 40 cycles of annea... View More
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Human Fractalkine ProQuantum Immunoassay Kit
          Human Fractalkine ProQuantum Immunoassay Kit
          Human Fractalkine ProQuantum Immunoassay Kit (A355825)
          videoThumbnail
          ►
          Product Specifications
          Analytical sensitivity
          <0.005 pg/mL
          Assay range
          0.0064-500 pg/mL
          Sample type/volume
          Serum
          2-5 µL
          Supernatant
          2-5 µL
          Plasma
          2-5 µL
          Hands-on time
          30 min
          Time-to-result
          2 hr
          Homogenous (no wash)
          Yes
          Instrument
          qPCR Instruments
          Product size
          5 x 96 Tests
          Contents
          Assay Dilution Buffer
          Antibody-Conjugate A
          Antibody-Conjugate B
          Antibody-Conjugate Dilution Buffer
          Standard
          Master Mix
          Ligase
          Storage
          -20°C
          Protein name
          CX3CL1
          Species (tested)
          Human
          Assay kit format
          Proximity-based Immunoassay
          Label or dye
          FAM™
          About This Kit
          The ProQuantum Human Fractalkine Immunoassay Kit is designed to provide highly sensitive quantitative measurements of Human Fractalkine protein in small sample volumes. Utilizing proximity ligation assay (PLA) technology, the assay combines the analyte specificity of high-affinity antibody-antigen binding with the signal detection and amplification capabilities of real-time PCR to achieve a simple yet powerful next-generation protein quantitation platform. A user-friendly workflow combined with intuitive Cloud-based software for analytics enables sample-to-answer in just 2 hours.
          • High sensitivity-detect low levels of protein with greater sensitivity than traditional methods
          • Broad dynamic range-≥5 logarithmic units, minimizing sample dilutions to ensure they fall within the range
          • Small sample consumption-use 2-5 µL of sample (compared to 75 µL for triplicate wells with other methods)
          • Fast, easy workflow-2 hours from sample to answer, with no wash steps
          • No proprietary instrument to purchase-runs on any real-time PCR instrument
          ProQuantum immunoassays utilize a matched pair of target-specific antibodies, each conjugated to a DNA oligonucleotide. During antibody-analyte binding, the two DNA oligos are brought into close proximity, which allows for ligation of the two strands and subsequent creation of a template strand for amplification. This platform leverages the sensitivity and large dynamic range of Applied Biosystems TaqMan real-time PCR technology (figure below).
          The assay workflow is fast and easy-2 steps in 2 hours. There are a total of 7 components in each kit (figure below). First, mix the antibody-conjugates, dilute the sample, and create the standard curve in a working plate. Then, using a multi-channel pipette, add the antibody-conjugates and sample (or standard) into the wells of a PCR plate and incubate for 1 hour. Combine the master mix and ligase and add to the wells of the PCR plate, then run the plate on any qPCR instrument. After the run is complete, import the results file into the ProQuantum cloud-based software at https://apps.thermofisher.com/apps/proquantum. Using this software, the data can be analyzed easily to obtain protein concentration values. The software allows you to set up standard curves, design plate layouts, set up customized assay instructions, and obtain robust statistical group-wise comparisons.
          Target information
          Chemokines are a family of proteins associated with the trafficking of leukocytes in immune surveillance and inflammatory cell recruitment. They are classified based on the positions of key cysteine residues. CX3CL1 is a CX3C chemokine known to induce adhesion and migration of leukocytes mediated by a membrane-bound and soluble form respectively. Recent experiments have shown that CX3CL1 can suppress the production of nitrous oxide, interleukin-6, and TNF-a in activated microglia and neuronal cells, suggesting that it may act as an intrinsic inhibitor against neurotoxicity by activated microglia. Its receptor, CX3CR1, also functions as a co-receptor for HIV-1 and HIV-2 envelope fusion and virus infection, which can be inhibited by CX3CL1.
          Bioinformatics
          Gene aliases : A-152E5.2, ABCD-3, C3Xkine, CX3CL1, CXC3, CXC3C, FKN, fractalkine, neurotactin, NTN, NTT, SCYD1
          Gene ID : (Human) 6376
          Gene symbol : CX3CL1
          Protein Aliases : C-X3-C motif chemokine 1, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CX3CL1, Fractalkine, Neurotactin, small inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, neurotactin), member-1, Small-inducible cytokine D1, unnamed protein product
          UniProt ID (Human) P78423
          Recommended Products
          IL-6 Human Matched Antibody Pair
          Because you are interested in IL-6 Human ELISA Kit.
          SIZE
          10 x 96 tests
          PRICE
          USD 400.00
          Cat # CHC1263
          guarantee_icon
          Performance Guarantee
          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline