Search Thermo Fisher Scientific
Analytical sensitivity
1.5 ng/mL
Assay range
1.64-400 ng/mL
Sample type/volume
Plasma
1 µL
Serum
1 µL
Supernatant
100 µL
Hands-on time
1 hr 20 min
Time-to-result
4 hr 45 min
Homogenous (no wash)
No
Interassay CV
<12%
Intraassay CV
<10%
Instrument
Colorimetric Microplate Reader
Product size
96 Tests
Contents
Pre-coated 96 well plate
Standard
Assay Diluent concentrate
Biotinylated Detection Antibody
SAV-HRP
Wash Buffer
Chromogen
Stop Solution
Adhesive Plate Covers
Shipping condition
Wet or Dry Ice
Storage
2-8°C
Protein name
APOE
Protein aliases
Apo-E, Apolipoprotein E, apolipoprotein E3
Protein family
GPCR (G-Protein Coupled Receptors)
Species (tested)
Human
Assay kit format
Sandwich ELISA Kit
Detector antibody conjugate
Biotin
Label or dye
HRP
Gene aliases
AD2, APO-E, APOE, LDLCQ5, LPG
Gene ID
Gene symbol
APOE
UniProt ID
The Human Apolipoprotein E (Hu ApoE) ELISA quantitates Hu Apoe in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ApoE.
Principle of the method
The Human ApoE solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Apo E (Apolipoprotein E) plays an important role in the metabolism of lipids in the plasma, and is also is a constituent of various plasma lipoprotein-lipid particles. The protein is synthesized in liver, brain, spleen, and the kidney, and concentrations are high in interstitial fluids, where it plays a role in distributing cholesterol from cells where cholesterol is in excess to cells which require cholesterol. Apo E also plays an important role in the formation of very low density lipoprotein and chylomicrons. The Apo E gene is located on chromosome 19 at position 19q13. 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support