  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • ELISA Kits ›
          • Fetuin B ELISA Kits
          Invitrogen
          Human FETUB/Fetuin B ELISA Kit
          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          Human FETUB/Fetuin B ELISA Kit
          Group 53 Created with Sketch.
          Human FETUB/Fetuin B ELISA Kit
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}
          Human FETUB/Fetuin B ELISA Kit
          ELISA Standard curve of FETUB/Fetuin B using Human FETUB/Fetuin B ELISA Kit (Product # EHFETUB). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Human FETUB/Fetuin B ELISA Kit
          Human FETUB/Fetuin B ELISA Kit (EHFETUB)
          videoThumbnail
          ►
          Product Specifications
          Analytical sensitivity
          4 ng/mL
          Assay range
          4.39-50 ng/mL
          Sample type/volume
          Plasma
          5 µL
          Serum
          5 µL
          Supernatant
          100 µL
          Hands-on time
          1 hr 20 min
          Time-to-result
          4 hr 45 min
          Homogenous (no wash)
          No
          Interassay CV
          <12%
          Intraassay CV
          <10%
          Instrument
          Absorbance Microplate Reader
          Product size
          96 Tests
          Contents
          Pre-coated 96 well plate
          Standard
          Assay Diluent concentrate
          Biotinylated Detection Antibody
          SAV-HRP
          Wash Buffer
          Chromogen
          Stop Solution
          Adhesive Plate Covers
          Shipping conditions
          Wet ice
          Storage
          Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
          Protein name
          Fetuin B
          Protein family
          Cell Structure/Organelle Proteins
          Species (tested)
          Human
          Assay kit format
          Sandwich ELISA Kit
          Detector antibody conjugate
          Biotin
          Label or dye
          HRP
          About This Kit
          The Human FETUB (Fetuin B) ELISA quantitates Hu FETUB in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu FETUB.
          Principle of the method
          The Human FETUB solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
          Rigorous validation
          Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
          Target information
          The protein encoded by this gene is a member of the fetuin family, part of the cystatin superfamily of cysteine protease inhibitors. Fetuins have been implicated in several diverse functions, including osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors, and response to systemic inflammation. This protein may be secreted by cells.
          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
          References (0)
          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          Bioinformatics
          Gene aliases : 16G2, FETUB, Gugu, IRL685
          Gene ID : (Human) 26998
          Gene symbol : FETUB
          Protein Aliases : 16G2, Fetuin-like protein IRL685, Gugu, Fetuin-B, Gugu beta, Gugu gamma, Gugu-a, unnamed protein product
          UniProt ID (Human) Q9UGM5
          Recommended Products
          IL-6 Human Matched Antibody Pair
          Because you are interested in IL-6 Human ELISA Kit.
          SIZE
          10 x 96 tests
          PRICE
          USD 400.00
          Cat # CHC1263
          guarantee_icon
          Performance Guarantee
          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
          Learn more
          help_icon
          We're here to help
          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline