Search Thermo Fisher Scientific
Analytical sensitivity
10 pg/mL
Assay range
12.35-3000 pg/mL
Sample type/volume
Plasma
5 µL
Serum
5 µL
Supernatant
50 µL
Hands-on time
1 hr 20 min
Time-to-result
4 hr 45 min
Homogenous (no wash)
No
Interassay CV
<12%
Intraassay CV
<10%
Instrument
Colorimetric Microplate Reader
Product size
96 Tests
Contents
Pre-coated 96 well plate
Standard
Assay Diluent concentrate
Biotinylated Detection Antibody
SAV-HRP
Wash Buffer
Chromogen
Stop Solution
Adhesive Plate Covers
Shipping condition
Wet or Dry Ice
Storage
2-8°C
Protein name
CD105 (Endoglin)
Protein aliases
CD105, CD105 antigen, Endoglin
Species (tested)
Human
Assay kit format
Sandwich ELISA Kit
Detector antibody conjugate
Biotin
Label or dye
HRP
Gene aliases
END, ENG, HHT1, ORW1
Gene ID
Gene symbol
ENG
UniProt ID
The Human Endoglin ELISA quantitates Hu Endoglin in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu endoglin.
Principle of the method
The Human Endoglin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support