Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Human CD46 ELISA Kit is an enzyme-linked immunosorbent assay for quantitative detection of human CD46 in serum, plasma, and cell culture supernatants.
CD46 (MCP, membrane cofactor protein) is a multifunctional cell surface transmembrane protein that binds and inactivates C3b and C4b complement fragments, regulates T cell-induced inflammatory responses by either inhibiting (CD46-1 isoform) or increasing (CD46-2 isoform) the contact hypersensitivity reaction. CD46 also serves as a receptor for several human pathogens (both bacteria and viruses), and its ligation alteres T lymphocyte polarization toward antigen-presenting cells or target cells, inhibiting lymphocyte function. CD46 is a protector of placental tissue and is also expressed on the inner acrosomal membrane of spermatozoa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : AHUS2, MCP, MIC10, TLX, TRA2.10
Gene ID : (Human) 4179
Gene symbol : CD46
Protein Aliases : antigen identified by monoclonal antibody TRA-2-10, CD46 antigen, complement regulatory protein, CD46 molecule, complement membrane cofactor protein, measles virus receptor, membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen), MGC26544, trophoblast leucocyte common antigen, trophoblast leukocyte common antigen, trophoblast-lymphocyte cross-reactive antigen, unnamed protein product
UniProt ID (Human) P15529
If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support