Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • ELISA Kits ›
          • PROK1 ELISA Kits
          This product (900-K244) is being sold under new product number 900-K244K.

          Invitrogen

          Human EG-VEGF/PROK1 ELISA Development Kit (ABTS), PeproTech®

          1 Reference
          View all (3) PROK1 ELISA Kits

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          Human EG-VEGF/PROK1 ELISA Development Kit (ABTS)
          Group 53 Created with Sketch.
          Human EG-VEGF/PROK1 ELISA Development Kit (ABTS)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Human EG-VEGF/PROK1 ELISA Development Kit (ABTS)

          EG-VEGF/PROK1 Human ABTS ELISA Standard Curve (Product # 900-K244). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Human EG-VEGF/PROK1 ELISA Development Kit (ABTS)
          Human EG-VEGF/PROK1 ELISA Development Kit (ABTS) (900-K244)
          videoThumbnail
          ►

          Product Specifications

          Analytical sensitivity

          Lot-specific

          Assay range

          Lot-specific

          Hands-on time

          1 hr 45 min

          Time-to-result

          24 hr 30 min

          Homogenous (no wash)

          No

          Instrument

          Absorbance Microplate Reader

          Product size

          10 x 96 Tests

          Contents

          This Human EG-VEGF ELISA Kit includes: Capture Antibody, Detection Antibody, Protein Standard, and HRP-Conjugate.

          Shipping conditions

          Ambient

          Storage

          Refer to product documentation for component specific storage temperature.

          Protein name

          PROK1

          Protein family

          Growth Factors & Receptors

          Species (tested)

          Human

          Assay kit format

          ELISA Development Kit

          Detector antibody conjugate

          Biotin

          Label or dye

          HRP

          About This Kit

          Human EG-VEGF ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human EG-VEGF in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human EG-VEGF in approximately 1,000 ELISA plate wells. This product is subject to certain licensed use limitations. For full details, please see product insert.

          This PeproTech ABTS ELISA Development Kit contains the key components required for the quantitative measurement of natural and/or recombinant proteins in a sandwich ELISA format. This ABTS ELISA Development Kit contains enough material to assay the target cytokine in at least 1,000 ELISA plate wells. The ABTS ELISA Buffer Kit (Product # 900-K00), sold separately, is recommended to be used in conjunction with this ABTS ELISA Development Kit for optimal performance. This buffer kit contains all the necessary components to assay ten 96-well ELISA plates (included) and contains detailed handling instructions.

          Target information

          Endocrine gland-derived vascular endothelial growth factor induces proliferation, migration, and fenestration in capillary endothelial cells derived from endocrine glands. Its expression is induced by hypoxia and is restricted to the steroidogenic glands. Its expression is often complementary to the expression of VEGF, suggesting that these molecules function in a coordinated manner.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Gene aliases : EGVEGF, PK1, PRK1, PROK1, UNQ600/PRO1186

          Gene ID : (Human) 84432

          Gene symbol : PROK1

          Protein Aliases : black mamba toxin-related protein, EG-VEGF, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, Prokineticin-1

          UniProt ID (Human) P58294

          Recommended Products

          IL-6 Human Matched Antibody Pair

          Because you are interested in IL-6 Human ELISA Kit.

          SIZE

          10 x 96 tests

          PRICE

          USD 400.00

          Cat # CHC1263

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline