Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • ELISA Kits ›
          • HLA-DR ELISA Kits

          Invitrogen

          Human HLA-DR ELISA Kit

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          Human HLA-DR ELISA Kit (EH550RB)
          Group 53 Created with Sketch.
          Human HLA-DR ELISA Kit (EH550RB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Human HLA-DR ELISA Kit

          Sandwich ELISA Kit {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Human HLA-DR ELISA Kit (EH550RB)
          videoThumbnail
          ►

          Product Specifications

          Analytical sensitivity

          1.76 ng/mL

          Assay range

          1.76-500 ng /mL

          Sample type/volume

          Plasma
          100 µL
          Supernatant
          100 µL
          Serum
          100 µL

          Hands-on time

          1 hr 20 min

          Time-to-result

          4 hr 45 min

          Homogenous (no wash)

          No

          Interassay CV

          12%

          Intraassay CV

          10%

          Instrument

          Absorbance Microplate Reader

          Product size

          96 Tests

          Contents

          Pre-coated microwell plate
          Standard
          Biotinylated Detection Antibody
          Streptavidin-HRP
          Sample Diluent
          Wash Buffer Concentrate
          Assay Buffer concentrate
          Stop Solution
          Substrate Solution
          Adhesive Plate Covers

          Shipping conditions

          Wet ice

          Storage

          -20°C

          Protein name

          HLA-DR

          Species (tested)

          Human

          Detector antibody conjugate

          Biotin

          Label or dye

          HRP

          About This Kit

          Human HLA-DR ELISA Kit is an enzyme-linked immunosorbent assay for quantitative detection of human HLA-DR.

          Target information

          HLA-DR, like other MHC class II molecules, is a transmembrane glycoprotein composed of a 36 kDa alpha chain (DRA) and 27 kDa beta chain (DRB). The alpha chain gene contains 5 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, and exon 4 encodes the transmembrane domain and the cytoplasmic tail. DRA does not have polymorphisms in the peptide binding part and acts as the sole alpha chain for DRB1, DRB3, DRB4 and DRB5. Within the DR molecule the beta chain contains all the polymorphisms specifying the peptide binding specificities. Hundreds of DRB1 alleles have been described and typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. HLA-DR is expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, thymic epithelial cells and activated T lymphocytes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides, present them to T-helper cells, and have a critical role in the initiation of the immune response.

          HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Gene aliases : CLIP, DHLAG, DR4, DRB1, DRB3, DRB4, DRB5, HLA DRB3, HLA-DR1B, HLA-DR3B, HLA-DR4B, HLA-DRA1, HLA-DRB, HLA-DRB3*, HLA-DRB4*, HLA-DRB5*, HLADG, Ia-GAMMA, II, p33, SS1

          Gene ID : (Human) 972, (Human) 3122, (Human) 3123, (Human) 3125, (Human) 3126, (Human) 3127

          Gene symbol : CD74, HLA-DRA, HLA-DRB1, HLA-DRB3, HLA-DRB4, HLA-DRB5

          Protein Aliases : CD74 antigen (invariant polypeptide of major histocompatibility complex, class II antigen-associated), CD74 antigen, DASS-397D15.1, FLJ51114, FLJ75017, FLJ76359, XXbac-BPG161M6.1, MHC2, MHCII, CD74 molecule, major histocompatibility complex, class II invariant chain, cell surface glycoprotein, integral membrane protein, class II HLA histocompatibility antigen, class II MHC-associated invariant chain peptide, DR beta-5, DR7 beta-chain glycoprotein, DRB1, DRB52, DRw53 beta-chain glycoprotein, ERLA-005, gamma chain of class II antigens, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR beta 3 chain, DR beta 4 chain, DR-1 beta chain, DR-5 beta chain, HLA-DR alpha-chain, hla-dr antigen alpha chain, hla-dr antigen heavy chain, HLA-DR antigens-associated invariant chain, HLA-DR-alpha chain precursor, HLA-DR-beta chain, HLA-DR-gamma, HLA-DR11-14N allele, HLA-DRA, HLA-DRB1, human leucocyte antigen, human leucocyte antigen DRB1, human leucocyte antigen DRB3, Ia-associated invariant chain, leukocyte antigen, lymphocyte antigen DRB1, class II, DRB3, MHC class II antigen, MHC class II antigen DR beta 3 chain, MHC class II antigen DRA, MHC class II antigen DRB3, MHC class II antigen DRB4, MHC class II antigen DRB5, MHC class II DR beta chain, MHC class II HLA-DR beta 1 chain, MHC class II HLA-DR beta 3 chain, MHC class II HLA-DR4 protein, MHC class II HLA-DR4/w53 protein, MHC class II protein, MHC DR beta protein precursor, MHC DR-beta, MHC DR7-beta-1 chain, MHC DR7-beta-2 chain, MHC HLA DR5 cell surface glycoprotein beta chain precursor, MHC HLA-DR gamma chain, MHC HLA-DR-beta chain, MHC HLA-DR-beta chain precursor, MHC HLA-DR-beta chain precursor old gene name 'HLA-DRA1', MHC HLA-DR-beta surface glycoprotein, MHC HLA-DR-beta-1 chain, MHC HLA-DR-beta-1 precursor, MHC HLA-DR-beta-2 precursor, MHC HLA-DR1-beta, MHC HLA-DR2 (non-Dw2/non-Dw12)b glycoprotein beta-chain, MHC HLA-DR2(Dw12)a glycoprotein beta-chain, MHC HLA-DR2(Dw12)b glycoprotein beta-chain, MHC HLA-DR2(Dw2)a glycoprotein beta-chain, MHC HLA-DR2(Dw2)b glycoprotein beta-chain, MHC HLA-DR2(non-Dw2/non-Dw12)a glycoprotein beta-chain, MHC HLA-DRw10-beta-2 protein precursor, unnamed protein product

          UniProt ID (Human) P04233, (Human) P01903, (Human) P01911, (Human) P79483, (Human) P13762, (Human) Q30154

          Recommended Products

          IL-6 Human Matched Antibody Pair

          Because you are interested in IL-6 Human ELISA Kit.

          SIZE

          10 x 96 tests

          PRICE

          USD 400.00

          Cat # CHC1263

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-svw7g:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch