Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The Human Interleukin 10 Receptor Alpha (CD210a) (Hu IL-10RA) ELISA quantitates Hu IL-10RA in human serum, plasma or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-10RA.
Principle of the method
The Human IL-10RA solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The protein encoded by this gene is a receptor for interleukin 10. This protein is structurally related to interferon receptors. It has been shown to mediate the immunosuppressive signal of interleukin 10, and thus inhibits the synthesis of proinflammatory cytokines. This receptor is reported to promote survival of progenitor myeloid cells through the insulin receptor substrate-2/PI 3-kinase/AKT pathway. Activation of this receptor leads to tyrosine phosphorylation of JAK1 and TYK2 kinases. Two transcript variants, one protein-coding and the other not protein-coding, have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : CD210, CD210a, CDW210A, HIL-10R, IL-10R1, IL10R
Gene ID : (Human) 3587
Gene symbol : IL10RA
Protein Aliases : CDW210B, CRF2-4, CRFB4, D21S58, D21S66, I10R, IL-10R, I10R1, IL-10R alpha, IL-10R2, il-10 receptor, il-10 receptor antagonist, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, interleukin-10 receptor subunit 1, unnamed protein product
UniProt ID (Human) Q13651
If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support