Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • ELISA Kits ›
          • Human IgE ELISA Kits

          Invitrogen

          Human IgE Uncoated ELISA Kit

          View all (4) Human IgE ELISA Kits

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          Human IgE Uncoated ELISA Kit (88-50610-88)
          Group 53 Created with Sketch.
          Human IgE Uncoated ELISA Kit (88-50610-88)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Human IgE Uncoated ELISA Kit

          Uncoated ELISA Kit {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Human IgE Uncoated ELISA Kit (88-50610-88)
          videoThumbnail
          ►

          Product Specifications

          Analytical sensitivity

          7.8 ng/mL

          Assay range

          7.8-500 ng/mL

          Sample type/volume

          Plasma
          10 µL
          Serum
          10 µL

          Hands-on time

          1 hr 45 min

          Time-to-result

          24 hr 30 min

          Homogenous (no wash)

          No

          Instrument

          Absorbance Microplate Reader

          Product size

          10 x 96 Tests

          Contents

          Capture Antibody: Pre-titrated, purified antibody
          Detection Antibody: Pre-titrated, HRP-conjugated antibody
          Standard: Human IgE protein for generating standard curve and calibrating samples
          10X Coating Buffer: Buffer for plating the Capture Antibody
          5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme
          Enzyme: Pre-titrated Avidin-HRP
          Substrate: 1X TMB Solution
          Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards
          96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)

          Shipping conditions

          Wet ice

          Storage

          2-8°C

          Protein name

          Human IgE

          Species (tested)

          Human

          Assay kit format

          Uncoated ELISA Kit

          Detector antibody conjugate

          Biotin

          Label or dye

          HRP

          About This Kit

          The Human Immunoglobulin E (Hu IgE) Uncoated ELISA Kit contains pre-matched antibody pairs and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IgE. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately.

          Principle of the method

          ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

          Target information

          IgE is one of the five classes of immunoglobulins found in mammals, characterized by its monomeric structure consisting of two heavy (epsilon) chains with four Ig-like constant domains and two light chains. Although IgE comprises only a small portion of the total immunoglobulin in serum, it plays a crucial role in type I hypersensitivity reactions, such as hay fever, asthma, hives, and anaphylactic shock. IgE is produced by terminally differentiated plasma B cells and basophils. It binds to Fc epsilon receptors on mast cells and basophils, leading to the release of histamine and other molecules that promote allergic responses. Crosslinking of IgE bound to these receptors induces degranulation of mast cells and basophils, resulting in allergic manifestations. In addition to its role in allergic responses, IgE is functionally important for immunity against parasites, highlighting its dual role in both protective and pathological immune processes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases : Immunoglobulin E, ImmunoglobulinE

          Recommended Products

          IL-6 Human Matched Antibody Pair

          Because you are interested in IL-6 Human ELISA Kit.

          SIZE

          10 x 96 tests

          PRICE

          USD 400.00

          Cat # CHC1263

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0