Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The Mouse TRACP/PAP/ACP5 Rapid ELISA Kit quantitates Tartrate Resistant Acid Phosphatase in serum and plasma
Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher's disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin's lymphoma and in the sera of patients undergoing active bone turnover.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : Acp5, T5ap, TRACP, TRAP
Gene ID : (Mouse) 11433
Gene symbol : Acp5
Protein Aliases : acid phosphatase type V, Tartrate-resistant acid ATPase, Tartrate-resistant acid phosphatase type 5, TR-AP, TRAPase, TrATPase, Type 5 acid phosphatase
UniProt ID (Mouse) Q05117
If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support