  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • ELISA Kits ›
          • Pregnanediol-3-Glucuronide ELISA Kits
          Invitrogen
          Pregnanediol-3-Glucuronide (PDG) Competitive ELISA Kit
          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          Pregnanediol-3-Glucuronide (PDG) Competitive ELISA Kit (EIAPDG)
          Group 53 Created with Sketch.
          Pregnanediol-3-Glucuronide (PDG) Competitive ELISA Kit (EIAPDG)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}
          Pregnanediol-3-Glucuronide (PDG) Competitive ELISA Kit
          Competitive ELISA {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Pregnanediol-3-Glucuronide (PDG) Competitive ELISA Kit (EIAPDG)
          videoThumbnail
          ►
          Product Specifications
          Analytical sensitivity
          0.18 ng/mL
          Assay range
          0.391-50 ng/mL
          Sample type/volume
          Fecal Extract
          50 µL
          Plasma
          50 µL
          Serum
          50 µL
          Supernatant
          50 µL
          Urine
          10 µL
          Hands-on time
          1 hr 20 min
          Time-to-result
          2 hr 30 min
          Homogenous (no wash)
          No
          Interassay CV
          6.4%
          Intraassay CV
          4.1%
          Instrument
          Absorbance Microplate Reader
          Product size
          96 Tests
          Contents
          Pre-coated 96 well plate
          Standard
          Assay Diluent concentrate
          HRP-Conjugated Detection Antibody
          Wash Buffer
          Chromogen
          Stop Solution
          Adhesive Plate Covers
          Shipping conditions
          Wet ice
          Storage
          -20°C
          Protein name
          Pregnanediol-3-Glucuronide
          Species (tested)
          Chemical
          Assay kit format
          Competitive ELISA
          Detector antibody conjugate
          HRP
          Label or dye
          HRP
          About This Kit
          The Pregnanediol-3-Glucuronide (PDG) ELISA quantitates PDG in extracted serum, plasma, urine, dried fecal extracts or cell culture medium.
          Principle of the method
          The Pregnanediol-3-Glucuronide (PDG) Competitive ELISA research-use-only kit uses a specifically generated antibody to measure PDG and its metabolites in urine and fecal samples, or in extracted serum and plasma. This kit is not recommended for serum, plasma, or saliva samples without extraction. The kit will quantitatively measure PDG present in diluted buffer samples and tissue culture media samples. A PDG standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture rabbit antibodies. A PDG-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a polyclonal antibody to PDG to each well. After a 2-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound PDG-peroxidase conjugate. After a short incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.
          Rigorous validation
          Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
          Target information
          Pregnanediol-3-Glucuronide (PDG) is a natural human metabolite of Pregnanediol generated in the liver by UDP glucuonyltransferase. Glucuronidation is used to assist in the excretion of toxic substances, drugs or other substances that cannot be used as an energy source. Glucuronic acid is attached via a glycosidic bond to the substance, and the resulting glucuronide, which has much higher water solubility than the original substance, is eventually excreted by the kidneys.
          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
          References (0)
          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          Bioinformatics
          Protein Aliases : PDG, C27H44O8
          Recommended Products
          IL-6 Human Matched Antibody Pair
          Because you are interested in IL-6 Human ELISA Kit.
          SIZE
          10 x 96 tests
          PRICE
          USD 400.00
          Cat # CHC1263
          guarantee_icon
          Performance Guarantee
          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
          Learn more
          help_icon
          We're here to help
          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline