Search
NucRed Dead 647 ReadyProbes Reagent emits bright far-red fluorescence when bound to DNA and is suitable for staining nuclei in fixed-cell preparations and tissue sections. NucRed Dead 647 reagent may also be used to measure viability and to measure cytotoxicity in kinetic live-cell assays.
First, fix and permeabilize cultured cells with a protocol appropriate for your sample.
| NucRed | |
|---|---|
| Excitation/Emission (nm) | 642/661 |
| Standard filter set | Cy5 |
| EVOS Light Cube | Cy5 |
| Storage conditions | Room temperature |
Tubulin and nuclear staining in HeLa cells. HeLa cells were fixed and permeabilized and stained for tubulin (biotinylated mouse anti-a-tubulin mouse monoclonal antibody followed by an Alexa Fluor 488 goat anti-mouse secondary antibody) and counter-stained with NucRed Dead 647 ReadyProbes reagent.
For Research Use Only. Not for use in diagnostic procedures.