Search
Automating microscope imaging improves your throughput and helps you analyze more samples. In different laboratory contexts, automation can address different steps in the analysis workflow. For some experiments, the goal is hands-off scanning for a microwell plate. Labs with higher throughput want to see robotic plate loading. Others want tools that will count neurite branches or intracellular vesicles in an automatic and reproducible way. Thermo Fisher Scientific has the tools for any level of automation.
See the entire portfolio of easy to use cell imaging instruments that can be configured with a variety of objectives, light cubes, and vessel holders to meet your research needs.
Thermo Fisher Scientific offers an automated imaging solution to fit your application—you choose the system that is right for you based on the imaging applications you want to run. Any or all of the following could influence your choice of platform:
Use the selection guide below to find a system that fits you.
![]() EVOS M7000 Perfect for a variety of advanced automated imaging applications | ![]() CellInsight CX5 platform Compact screening system to scale up your throughput | |
| Widefield fluorescence | 4-channel | 5-channel |
| Camera | High-sensitivity 3.2 MP (2048 x 1536) CMOS sensor with 3.45 µm pixel resolution | Photometrics X1 CCD |
| LED light cubes | ✓ | |
| LED light engine | ✓ | |
| Fluorescence channels | Simultaneously accommodates 4 interchangeable fluorescence cubes plus bright-field imaging. More than 20 light cubes to choose from. | Blue Ex: 386/23 Em: 440/40 Green Ex: 485/20 Em: 521/21 Red Ex: 560/25 Em: 607/34 Far Red Ex: 650/13 Em: 694/35 Near IR Ex:740/13 Em: 809/81 |
| Confocal | NA | NA |
| Brightfield | ✓ | ✓ |
| Phase contrast | ✓ | |
| Objective turret | 5-position 1.25x–100x | 1-position 2x–20x |
| Autofocus | ✓ | ✓ |
| Onstage incubator | ✓ (optional) | |
| Automated scanning | ✓ | ✓ |
| Robot compatible | ✓ | |
| Software | EVOS M7000 with optional Celleste Image Analysis Software | HCS Studio |
| Weight | 11.8 kg (26 lbs) | 45 kg (99.2 lb) |
| Dimensions | H: 36 cm (13") W: 33 cm (14") D: 46 cm (18") | H: 37.2 cm (14.6") W: 41.4 cm (16.3") D: 59.4 cm (23.4") |
Cell Analysis Support Center
Find tips, troubleshooting help, and resources for your cell analysis workflow.
For Research Use Only. Not for use in diagnostic procedures.