Measuring a cell’s ability to proliferate is fundamental for assessing cell health, determining genotoxicity, and evaluating anti-cancer drugs. The most accurate method is by directly measuring DNA synthesis. Initially, radioactive nucleosides like 3H-thymidine were used, later replaced by antibody-based detection of bromo-deoxyuridine (BrdU). The Click-iT EdU flow cytometry assays offer a novel alternative using EdU (5-ethynyl-2´-deoxyuridine), which incorporates into DNA synthesis is detected via a copper-catalyzed click reaction between an azide and an alkyne. The azide is coupled to dyes such as Pacific Blue, Alexa Fluor 647, or Alexa Fluor 488. Then, standard flow cytometry methods are used to determine the percentage of S-phase cells in the population. Click-iT EdU labeling is advantageous due to the small size of the dye azide, allowing efficient detection under mild conditions, unlike BrdU assays that require DNA denaturation, which can alter cell cycle distribution and destroy antigen recognition sites. The EdU assay is compatible with cell cycle dyes and can be multiplexed with antibodies against surface and intracellular markers, though some reagents may require additional steps for compatibility.
See all Click-iT Edu cell proliferation assays
Materials provided in kit
| Material | Amount | Concentration |
|---|
| EdU (Component A) | 10 mg | NA |
| Alexa Fluor 488 azide, Alexa Fluor 647 azide, or Pacific Blue azide (Component B) | 1 vial | NA |
| Dimethylsulfoxide (DMSO) (Component C) | 4.25 mL | NA |
| Click-iT fixative (Component D) | 5 mL | 4% paraformaldehyde in PBS |
| Click-iT saponin-based permeabilization and wash reagent (Component E) | 50 mL | 10X solution |
| CuSO4 (Component F) | 0.5 mL | 100 mM aqueous solution |
| Click-iT EdU buffer additive (Component G) | 400 mg | NA |
| Number of assays: Sufficient material is supplied for 50 reactions, based on the protocol below. |
| Approximate fluorescence excitation/emission maxima: Pacific Blue azide: 410/455 nm; Alexa Fluor 647 azide: 650/670 nm; Alexa Fluor 488 azide: 495/519 nm. |
Materials required but not provided
- 1% bovine serum albumin (BSA) in phosphate buffered saline (PBS), pH 7.1–pH 7.4
- Buffered saline solution, such as PBS, D-PBS, or TBS
- Deionized water or 18 MΩ purified water
- 12 × 75-mm tubes, or other flow cytometry tubes
Cautions
- DMSO (Component C), provided as a solvent in this kit, is known to facilitate the entry of organic molecules into tissues. Handle reagents containing DMSO using equipment and practices appropriate for the hazards posed by such materials. Dispose of the reagents in compliance with all pertaining local regulations.
- Click-iT fixative (Component D) contains paraformaldehyde, which is harmful. Use with appropriate precautions.
- Click-iT saponin-based permeabilization and wash reagent (Component E) contains sodium azide, which yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.
Top
Protocol steps
Preparing reagents
- Allow vials to warm to room temperature before opening.
- To prepare a 10 mM solution of EdU, add 4 mL of DMSO (Component C) or aqueous solution (PBS) to Component A and mix well. After use, store any remaining stock solution at ≤–20°C. When stored as directed, the stock solution is stable for up to 1 year.
- To prepare a working solution of Pacific Blue azide, Alexa Fluor 647 azide, or Alexa Fluor 488 azide, add 130 μL of DMSO to Component B and mix well. After use, store any remaining working solution at ≤–20°C. When stored as directed, this working solution is stable for up to 1 year.
- To prepare 500 mL of 1X Click-iT saponin-based permeabilization and wash reagent, add 50 mL of Component E to 450 mL of 1% BSA in PBS. Smaller amounts can be prepared by diluting a volume of Component E 1:10 with 1% BSA in PBS. After use, store any remaining solutions at 2–6˚C. When stored as directed, the 1X solution is stable for 6 months and the 10X solution is stable for 12 months after receipt. Note: Component E contains sodium azide (see Cautions).
- To make a 10X stock solution of the Click-iT EdU buffer additive (Component G), add 2 mL of deionized water to the vial and mix until the Click-iT EdU buffer additive is fully dissolved. After use, store any remaining stock solution at ≤–20˚C. When stored as directed, the stock solution is stable for up to 1 year.
Labeling cells with EdU
The following protocol was developed with Jurkat cells, a human T cell line, and using an EdU concentration of 10 μM, and can be adapted for any cell type. Growth medium, cell density, cell type variations, and other factors may influence labeling. In initial experiments, we recommend testing a range of EdU concentrations to determine the optimal concentration for your cell type and experimental conditions. If currently using a BrdU based assay for cell proliferation, a similar concentration to BrdU is a good starting concentration for EdU. If using whole blood as the sample, we recommend heparin as the anticoagulant for collection.
- Suspend the cells in an appropriate tissue culture medium to obtain optimal conditions for cell growth. Disturbing the cells by temperature changes or washing prior to incubation with EdU slows the growth of the cells during incorporation.
- Add EdU to the culture medium at the desired final concentration and mix well. We recommend a starting concentration of 10 μM for 1–2 hours. For longer incubations, use lower concentrations. For shorter incubations, higher concentrations may be required. For a negative staining control, include cells from the same population that have not been treated with EdU.
- Incubate under conditions optimal for cell type for the desired length of time. Altering the amount of time the cells are exposed to EdU or subjecting the cells to pulse labeling with EdU allows the evaluation of various DNA synthesis and proliferation parameters. Effective time intervals for pulse labeling and the length of each pulse depend on the cell growth rate.
- Harvest cells and proceed immediately to Staining cell-surface antigens with antibodies if performing antibody surface labeling; otherwise continue to Fixation and permeabilization.
Staining cell-surface antigens with antibodies (optional)
- Wash cells once with 3 mL of 1% BSA in PBS, pellet cells by centrifugation, and remove supernatant.
- Dislodge the pellet and resuspend cells at 1 × 107 cells/mL in 1% BSA in PBS.
- Add 100 μL of cell suspension or whole blood sample to flow tubes.
- Add surface antibodies and mix well (Table 2). Note: Do not use PE, PE-tandem, or Qdot antibody conjugates before performing the click reaction; wait until step 6.1 for labeling with these fluorophores.
- Incubate for the recommended time and temperature, protected from light. 3.6 Proceed to step 4.1 for cell fixation.
Fixation and permeabilization
The Click-iT saponin-based permeabilization and wash reagent can be used with whole blood or cell suspensions containing red blood cells, as well as with cell suspensions containing more than one cell type. This permeabilization and wash reagent maintains the morphological light scatter characteristics of leukocytes while lysing red blood cells.
- Wash the cells once with 3 mL of 1% BSA in PBS, pellet the cells, and remove the supernatant.
- Dislodge the pellet, add 100 μL of Click-iT fixative (Component D), and mix well.
- Incubate the cells for 15 minutes at room temperature, protected from light.
- Wash the cells with 3 mL of 1% BSA in PBS, pellet the cells, and remove the supernatant. Repeat the wash step if red blood cells or hemoglobin are present in the sample. Remove all residual red blood cell debris and hemoglobin before proceeding.
- Dislodge the cell pellet and resuspend the cells in 100 μL of 1X Click-iT saponin-based permeabilization and wash reagent (prepared in step 4 under Preparing reagents), and mix well. Incubate the cells for 15 minutes or proceed directly to the next protocol for click labeling.
Click-iT reaction
- Prepare 1X Click-iT EdU buffer additive by diluting the 10X stock solution (prepared in step 5 under Preparing reagents) 1:10 in deionized water.
- Prepare the Click-iT reaction cocktail according to Table 1.
NOTE: Use the Click-iT reaction cocktail within 15 minutes of preparation.
Table 1. Click-iT EdU reaction cocktails.
| Reaction components | Number of reactions |
|---|
| 1 | 2 | 5 | 10 | 15 | 30 | 50 |
|---|
| PBS, D-PBS, or TBS | 438 µL | 875 µL | 2.19 mL | 4.38 mL | 6.57 mL | 13.2 mL | 21.9 mL |
| CuSO4 (Component F) | 10 µL | 20 µL | 50 µL | 100 µL | 150 µL | 300 µL | 500 µL |
| Fluorescent dye azide | 2.5 µL | 5 µL | 12.5 µL | 25 µL | 37.5 µL | 75 µL | 125 µL |
| Reaction Buffer Additive | 50 µL | 100 µL | 250 µL | 500 µL | 750 µL | 1.5 mL | 2.5 mL |
| Total reaction volume | 500 µL | 1 mL | 2.5 mL | 5 mL | 7.5 mL | 15 mL | 25 mL |
- Add 0.5 mL of Click-iT reaction cocktail to each tube and mix well.
- Incubate the reaction mixture for 30 minutes at room temperature, protected from light.
- Wash the cells once with 3 mL of 1X Click-iT saponin-based permeabilization and wash reagent (prepared in step 4 under Preparing reagents), pellet the cells, and remove the supernatant. Dislodge the cell pellet and resuspend the cells in 100 μL of 1X Click-iT saponin-based permeabilization and wash reagent, if proceeding with intracellular antibody labeling in Staining cells for DNA content. Otherwise, add 500 μL of 1X Click-iT saponin-based permeabilization and wash reagent and proceed to Analysis by flow cytometry.
Staining cells for DNA content (optional)
- If necessary, add Ribonuclease A to each tube and mix (Table 2).
- Add the appropriate DNA stain to each tube, mix well, and incubate as recommended for each DNA stain.
Table 2. Click-iT EdU compatibility with DNA content stains.
| DNA content stain | Click-iT EdU stain compatibility | RNase required? |
|---|
| Pacific Blue | Alexa Fluor 647 | Alexa Fluor 488 |
|---|
| FxCycle Violet | No | Yes | Yes | No |
| Propidium iodide (PI) | Yes | Yes | No* | Yes |
| SYTOX AADvanced | Yes | Yes | Yes | Yes |
| FxCycle Far Red | Yes | No | Yes | Yes |
Analysis by flow cytometry
If measuring total DNA content on a traditional flow cytometer using hydrodynamic focusing, use a low flow rate during acquisition. If using the Attune Acoustic Focusing Cytometer, all collection rates may be used without loss of signal integrity if the event rate is kept below 10,000 events per second. However, for each sample within an experiment, the same collection rate and cell concentration should be used. The fluorescent signal generated by DNA content stains is best detected with linear amplification. The fluorescent signal generated by Click-iT EdU labeling is best detected with logarithmic amplification.
- Analyze the cells using a flow cytometer.
- For the detection of EdU with Pacific Blue azide, use 405 nm excitation with a violet emission filter (450/50 nm or similar).
- For the detection of EdU with Alexa Fluor 647 azide use 633/635 nm excitation with a red emission filter (660/20 nm or similar).
- For the detection of EdU with Alexa Fluor 488 azide, use 488 nm excitation with a green emission filter (530/30 nm or similar).
Ordering information
C10418,C10424,C10425,C10419,C10420,A10044,F10347,F10348,H3570,P3566,S10349,V35003,14190144,14190250
Frequently asked questions
What is the difference between BrdU and EdU assay?
The BrdU assay requires DNA denaturation and antibody staining to detect incorporated BrdU, while the EdU assay uses click chemistry for detection, eliminating the need for DNA denaturation. EdU offers benefits over BrdU, including a faster, simpler process and higher sensitivity.
How does EdU labeling work?
EdU labeling works by incorporating 5-ethynyl-2'-deoxyuridine (EdU), a thymidine analog, into newly synthesized DNA during the S-phase of the cell cycle. After incorporation, the EdU is detected using a copper-catalyzed click chemistry reaction with a fluorescent azide, which binds to the EdU. This method allows for the visualization and quantification of cell proliferation without the need for DNA denaturation, making the process faster and less harsh on cells.
What are EdU positive cells?
EdU positive cells are cells that have incorporated 5-ethynyl-2'-deoxyuridine (EdU) into their DNA during the S-phase of the cell cycle. These cells are actively proliferating and can be detected using click chemistry, which binds a fluorescent azide to the incorporated EdU, allowing for visualization and quantification of cell proliferation.
How does EdU Click-iT work?
EdU Click-iT works by incorporating EdU into newly synthesized DNA during cell proliferation. Detection involves a copper-catalyzed click chemistry reaction, where a fluorescent azide binds to the EdU. This reaction is fast and does not require DNA denaturation, allowing for easy visualization and quantification of proliferating cells.
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments