Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Amino Acids | |||
| Glycine | 75.0 | 500.0 | 6.6666665 |
| L-Alanine | 89.0 | 250.0 | 2.8089888 |
| L-Arginine hydrochloride | 211.0 | 700.0 | 3.3175356 |
| L-Aspartic acid | 133.0 | 300.0 | 2.255639 |
| L-Cysteine hydrochloride-H2O | 176.0 | 1.1 | 0.00625 |
| L-Cystine 2HCl | 313.0 | 260.0 | 0.83067095 |
| L-Glutamic Acid-H2O | 165.0 | 750.0 | 4.5454545 |
| L-Histidine hydrochloride-H2O | 210.0 | 218.8 | 1.0419048 |
| L-Hydroxyproline | 131.0 | 100.0 | 0.7633588 |
| L-Isoleucine | 131.0 | 400.0 | 3.053435 |
| L-Leucine | 131.0 | 600.0 | 4.5801525 |
| L-Lysine hydrochloride | 183.0 | 700.0 | 3.8251367 |
| L-Methionine | 149.0 | 150.0 | 1.0067114 |
| L-Phenylalanine | 165.0 | 250.0 | 1.5151515 |
| L-Proline | 115.0 | 400.0 | 3.4782608 |
| L-Serine | 105.0 | 250.0 | 2.3809524 |
| L-Threonine | 119.0 | 300.0 | 2.5210085 |
| L-Tryptophan | 204.0 | 100.0 | 0.49019608 |
| L-Tyrosine disodium salt dihydrate | 261.0 | 580.0 | 2.2222223 |
| L-Valine | 117.0 | 250.0 | 2.1367521 |
| Vitamins | |||
| Ascorbic Acid | 176.0 | 0.5 | 0.0028409092 |
| Biotin | 244.0 | 0.1 | 4.0983607E-4 |
| Choline chloride | 140.0 | 5.0 | 0.035714287 |
| D-Calcium pantothenate | 477.0 | 0.1 | 2.0964361E-4 |
| Folic Acid | 441.0 | 0.1 | 2.2675737E-4 |
| Menadione (Vitamin K3) | 172.0 | 0.1 | 5.8139535E-4 |
| Niacinamide | 122.0 | 0.25 | 0.0020491802 |
| Nicotinic acid (Niacin) | 123.0 | 0.25 | 0.0020325202 |
| Para-Aminobenzoic Acid | 137.0 | 0.5 | 0.003649635 |
| Pyridoxal hydrochloride | 204.0 | 0.25 | 0.0012254902 |
| Pyridoxine hydrochloride | 206.0 | 0.25 | 0.0012135922 |
| Riboflavin | 376.0 | 0.1 | 2.6595744E-4 |
| Thiamine hydrochloride | 337.0 | 0.1 | 2.967359E-4 |
| Vitamin A (acetate) | 328.0 | 1.0 | 0.0030487804 |
| Vitamin D2 (Calciferol) | 397.0 | 1.0 | 0.0025188916 |
| alpha Tocopherol phos. Na salt | 554.7 | 0.1 | 1.8027762E-4 |
| i-Inositol | 180.0 | 0.5 | 0.0027777778 |
| Inorganic Salts | |||
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 2000.0 | 18.018019 |
| Ferric nitrate (Fe(NO3)-9H2O) | 404.0 | 7.2 | 0.017821781 |
| Magnesium Sulfate (MgSO4) (anhyd.) | 120.0 | 970.0 | 8.083333 |
| Potassium Chloride (KCl) | 75.0 | 4000.0 | 53.333332 |
| Sodium Chloride (NaCl) | 58.0 | 68000.0 | 1172.4138 |
| Sodium Phosphate monobasic (NaH2PO4) anhydrous | 138.0 | 1400.0 | 10.144928 |
| Other Components | |||
| 2-deoxy-D-ribose | 134.0 | 5.0 | 0.03731343 |
| Adenine sulfate | 404.0 | 100.0 | 0.24752475 |
| Adenosine 5'-phosphate | 347.0 | 2.0 | 0.0057636886 |
| Adenosine 5'-triphosphate | 605.0 | 10.0 | 0.016528925 |
| Cholesterol | 387.0 | 2.0 | 0.005167959 |
| D-Glucose (Dextrose) | 180.0 | 10000.0 | 55.555557 |
| Glutathione (reduced) | 307.0 | 0.5 | 0.0016286644 |
| Guanine hydrochloride | 188.0 | 3.0 | 0.015957447 |
| Hypoxanthine Na | 158.0 | 3.5 | 0.022151899 |
| Phenol Red | 376.4 | 200.0 | 0.53134966 |
| Ribose | 150.0 | 5.0 | 0.033333335 |
| Sodium Acetate | 82.0 | 500.0 | 6.097561 |
| Thymine | 126.0 | 3.0 | 0.023809524 |
| Tween 80® | 200.0 | Infinity | |
| Uracil | 112.0 | 3.0 | 0.026785715 |
| Xanthine-Na | 152.0 | 3.44 | 0.02263158 |
1. Morgan, J.F. and Campbell, M.E. (1955) J. Natl. Cancer Inst., 16:557.
2. Morgan, J.F., Morton, H.J. and Parker R.C. (1950) Proc. Soc. Exp. Biol. Med., 73:1.
12340 - Medium 199, HEPES
Cat No:
12340030
11043 - Medium 199, no phenol red
Cat No:
11043023
22340 - Medium 199, HEPES
Cat No:
22340020
,
22340087
21180 - Medium 199, 10x
Cat No:
21180021
41150 - Medium 199, GlutaMAX(TM)
Cat No:
41150020
,
41150087
31153 - Medium 199, low sodium bicarbonate
Cat No:
31153026