Search
All EVOS imaging systems deliver advanced capabilities designed to make high-quality imaging faster, simpler, and more intuitive. From powerful software with an intuitive user interface to flexible magnification and illumination to easy access to data, EVOS imaging systems empower researchers to capture publication-quality images with ease and efficiency.
EVOS "M" |
EVOS "S" |
||
|---|---|---|---|
![]() EVOS M3000 |
![]() EVOS M5000 |
![]() EVOS M7000 |
![]() EVOS S1000 |
| Sample | |||
| Cell & tissue | Cell & tissue | Cell & tissue | Tissue |
| Maximum number of fluorescent colors | |||
| 2 | 4 | 4 | 9 |
| Automated image acquisition | |||
| No | No | Yes | Yes |
Generating clear and detailed images is crucial for outstanding visualization and analysis of cellular structures and processes. EVOS imaging systems are engineered for remarkable image quality. In a few clicks, you can capture bright, publication-quality images and videos that effectively showcase your data.
EVOS imaging systems offer quick setup and intuitive controls for straightforward operation with minimal training. All components and controls are conveniently integrated into a single, compact system that makes image capture easy, allowing you to go from sample to results faster. Large, bright monitors allow you to view images outside a darkroom and facilities safe collaboration in any space.
With the automated EVOS imaging systems, EVOS M7000 Imaging System and EVOS S1000 Spatial Imaging System, software controls most or all microscope operations. You can adjust focus, zoom, pan, and even control stage movement remotely for minimal handling.
All EVOS imaging systems include onboard software that enables users to effortlessly view, capture, and process images, with simple controls for adjusting colors, brightness, contrast, and more. The EVOS M7000 Imaging System expands these capabilities with advanced tools for cell counting, Z-stacking, image tiling, and time-lapse imaging, all accessible through an intuitive interface that requires minimal training.
Optional image analysis software is available for more advanced analysis, including classification, segmentation, and 3D visualization, helping to ensure that even the most complex workflows are simplified and efficient.
All EVOS imaging systems feature high-performance objectives lenses that span a full range of magnifications and specifications. The EVOS "M" microscopes also feature bright, interchangeable LED light cubes that span the entire spectrum while the EVOS S1000 system is powered by a custom LED engine. All can be customized to support a wide range of applications—from routine cell culture imaging to multiplex spatial imaging.
EVOS imaging systems are designed for collaboration and data management, offering networking capabilities that facilitate easy and secure data transfer, long-term data storage, and easy sharing. The ability to connect to a shared network and export captured images helps provide quick and easy access to data, from anywhere at any time.
For Research Use Only. Not for use in diagnostic procedures.