Search
Discover the unparalleled precision and efficiency of Thermo Scientific Multidrop reagent dispensers, designed to revolutionize your laboratory workflows. These cutting-edge instruments are packed with features that cater to the diverse needs of modern labs, from ensuring consistent, reproducible results to significantly reducing reagent costs. With an extensive volume range, Multidrop dispensers offer unmatched versatility, making them suitable for a wide array of applications, from low-volume qPCR to complex cell-based assays. Whether you’re looking to enhance your productivity, reduce waste, or simply achieve more accurate results, the Multidrop reagent dispensers are engineered to meet your demands. Explore the key features that make these dispensers an indispensable tool in laboratories worldwide.
In today's fast-paced laboratory environments, precision and reproducibility are paramount to ensuring reliable results and maximizing productivity. Thermo Scientific™ Multidrop™ reagent dispensers are engineered to meet these demands, offering advanced liquid handling solutions that cater to a wide range of applications. Whether your focus is on high-throughput screening, low-volume assays, or complex protocol automation, the Multidrop™ series provides the accuracy and consistency necessary to drive meaningful outcomes in your research. Below, we explore the precision and reproducibility features of the Multidrop Combi+, Multidrop Combi nL, and Multidrop Pico dispensers, highlighting how each instrument is tailored to meet the specific needs of modern laboratories.
Multidrop Combi+
Designed for high precision and reproducibility, making it an essential tool for laboratories that require accurate liquid handling. Utilizing advanced peristaltic pump technology and specialized dispensing cassettes, the Combi+ ensures precise dispensing across a broad volume range, from 0.5 µL to 2,500 µL. The ability to adjust volumes for each individual column and backflush to recover reagents further enhances the accuracy and efficiency of your workflow, minimizing waste and maximizing productivity.
Multidrop Combi nL
Unparalleled precision in low-volume dispensing, making it ideal for pharmaceutical and biotechnology laboratories focused on high-throughput, low-volume assays. Capable of dispensing volumes as small as 50 nL, the Combi nL employs valve-based dispensing technology to deliver consistent and accurate results across various plate formats.
Multidrop Pico
Redefine precision in digital dispensing, offering exceptional accuracy with volumes ranging from as low as 11 pL to 10 µL. These dispensers are particularly suited for applications requiring minimal reagent use, such as assay miniaturization and drug screening assays.
In the ever-evolving landscape of laboratory research, versatility is key to addressing diverse experimental requirements and adapting to various workflows. Thermo Scientific Multidrop family of reagent dispensers designed with this principle in mind, offering unparalleled flexibility to meet the unique demands of modern laboratories. Whether you’re working with a wide range of plate formats, accommodating different fluid types, or requiring precise dispensing across various volumes, Multidrop dispensers provide the adaptability needed to streamline operations and enhance productivity.
Despite their advanced capabilities, Multidrop dispensers are incredibly user-friendly, with intuitive interfaces that simplify setup and operation. Researchers can easily customize dispensing protocols to meet the specific needs of their experiments, from adjusting volumes and fluid types to selecting specific wells for dispensing. This ease of use, coupled with the ability to store and recall customized protocols, makes the Multidrop™ dispensers an ideal choice for laboratories of all sizes and specializations.
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.