image of Molecular Probes® NucGreen® Dead 488 ReadyProbes® Reagent
Nuclear stain for fixed cells
NucGreen Dead 488 ReadyProbes Reagent emits bright green fluorescence when bound to DNA and is suitable for staining nuclei in fixed-cell preparations and tissue sections. NucGreen Dead 488 reagent may also be used to measure viability and to measure cytotoxicity in kinetic live-cell assays. 
This protocol can be used for:
  • Nucleic acid (nuclear) staining in fluorescence microscopy
This protocol should not be used for:
  • Flow cytometry
You will need the following for this protocol:
Protocol
Labeling fixed cells
First, fix and permeabilize cultured cells with a protocol appropriate for your sample.
1. Wash the cells 1–3 times in PBS as needed.
2. Add two drops of NucGreen Dead 488 Reagent per milliliter of medium.
3. Incubate for 5–15 minutes, protected from light.
4. Remove the stain solution.
5. Wash the cells 2–3 times in PBS.
6. Image the cells.
Spectral information and storage
 NucGreen Dead 488
Excitation/Emission (nm)504/523  
Standard filter setFITC or GFP
EVOS Light CubeFITC
Storage conditionsRoom temperature
Protocol tips
HeLa cells stained with NucGreen® Dead 488 ReadyProbes®
HeLa cells stained with NucGreen Dead 488 ReadyProbes reagent, indicating loss of plasma membrane integrity.
Ordering information
Catalog # Name Size Price (USD) Qty
R37109 NucGreen™ Dead 488 ReadyProbes™ Reagent (SYTOX™ Green) Each
317.65
Online Exclusive
352.00
Save 34.35 (10%)