On this page:
Proven bioprocess technology to support your process
The HyPerforma Single-Use Bioreactor (S.U.B.) was designed for high performance, ease-of-use, and high-yield mammalian cell culture and microcarrier process results—all in a flexible solution that can be customized to your workflow and desired end goals.
Key performance of HyPerforma Single-Use Bioreactor
  • Ability to readily scale from small to large volumes—use in process development, clinical trials, and large-scale cGMP biomanufacturing
  • Plug-and-play setup and operation through standard disposable connectors, facilitating easy integration with most control platforms on the market
  • Minimized cleaning or sterilization (apart from traditional monitoring probes); helps reduce the risk of cross-contamination
  • Standard hardware and BPC options, including enhanced application specific designs
HyPerforma bioreactor 5:1 turndown ratio vessel
The HyPerforma S.U.B. helps decrease additional bioreactors required for scale-up, as well as reduce footprint and cleanroom space requirements. Important features include:
  • Patented crossflow sparger technology for efficient cell culture at lower volumes
  • Drilled-hole sparger for effective stripping of CO2 and sufficient O2 delivery to cells
  • Lower probe cutouts on vessel for accurate sensing of pH, DO, and temperature
  • Bottom jacketed vessel for better heat transfer and temperature control at lower volumes
  • Considerable savings in CAPEX and OPEX
Please note that the HyPerforma S.U.B. is also available with a 2:1 turndown ratio. 
Bioprocessing containers (BPCs) specific for single-use bioreactor systems
The single-use bioreactor bag—featuring the BioTitan Retention device—is the single-use consumable bag required for use with the bioreactor hardware; it is supplied sterile and ready-to-use and comes complete with the agitator (impeller) assembly, spargers, vent filter, sensor ports and/or sensors, and line sets for media, supplements, and harvest.
The single-use bioreactor bag does not require any facility hook-ups for sterilization or cleaning. A key element of the design is the single-use impeller with bearing/seal assembly that couples directly to the external mixer drive.
The Thermo Scientific Aegis and CX Film portfolio offer a complete bioreactor solution—from process development to commercial production while also helping you reach your sustainability goals in lowering emissions.
Interested in sustainability in bioprocessing? Enjoy the same robust film but now with carbon reduction. Learn more about Thermo Scientific Sustainable film.
HyPerforma Bioreactor automation software and controllers
  • Our HyPerforma Bioreactor portfolio can fully integrate with the Delta V-based HyPerforma G3 Bioprocess Controller systems that offer immediate utility.
  • Easily scale up and transfer your data. The G3 Bioprocess Controller systems consist of a range of controllers that can be used in research, process development, and full commercial manufacturing.
  • Thermo Scientific TruBio software facilitates easy, reliable, and repeatable process development and commercial cell culture processes.
HyPerforma Single-Use Bioreactor literature
Brochure: HyPerforma 5:1 Single-Use Bioreactor for optimal cell culture performance                                                        
Application note: S.U.B. enhancements for high-density perfusion cultures
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.