Search
From design to synthesis, quality primers are vital to successful results. Search our tool, to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and primer-dimers, highly target-specific, and used under universal PCR conditions - these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All are checked by mass spectometry. Have confidence knowing primer pairs are selected to tile across exons for optimal exon coverage with a minimal number of primer pairs.
Save time with the Primer Designer™ Tool by selecting from >300,000 primer pairs targeting the human exome.
For Research Use Only. Not for use in diagnostic procedures.