Image of SYTO® 82 Orange Nucleic Acid Stain
Nuclear stain for eukaryotic and prokaryotic cells
SYTO 82 is a cell-permeant, non-exclusive nucleic acid stain that shows a large fluorescence enhancement upon binding. In both live and dead eukaryotic cells, SYTO 82 generally shows cytoplasmic or mitochondrial as well as nuclear staining. In addition, SYTO 82 will stain most live and permeabilized bacteria.
This protocol can be used for:
  • Nucleic acid (nuclear) staining in fluorescence microscopy
This protocol should not be used for:
  • Flow cytometry
You will need the following for this protocol:
Protocol
1. Culture cells in an appropriate medium and vessel for fluorescence microscopy.
2. Remove the medium.
3. Wash the cells 1–3 times in a phosphate-free buffer to remove the medium.
4. Prepare the SYTO 82 staining solution by diluting the stock solution 1:1,000 (5 µM) in a phosphate-free buffer.
5. Add sufficient staining solution to cover the cells.
6. Incubate for 5–30 minutes, protected from light.
7. Remove the staining solution.
8. Wash sample 3 times in a phosphate-free buffer.
9. Image the cells.
Spectral information and storage
 SYTO 82
Excitation/Emission (nm)541/560
Standard filter setTRITC
EVOS Light CubeRFP
Storage conditions≤–20°C
Protocol tips
  • Warm to room temperature and briefly centrifuge the DMSO solution to the bottom of the vial each time before use.
  • Try multiple dye concentrations in the range from 100 nM to 5 µM to determine the optimal concentration.
  • In general, the best results are obtained in buffers that do not contain phosphate, such as Hank’s Balanced Salt Solution (Cat. No. 14025092).
Cells stained with SYTO® 82
Cells stained with SYTO 82 dye and imaged with the Thermo Scientific CellInsight High-Content System.
Ordering information
Catalog # Name Size Price (USD) Qty
S11363 SYTO™ 82 Orange Fluorescent Nucleic Acid Stain - 5 mM Solution in DMSO Each
363.65
Online Exclusive
376.00
Save 12.35 (3%)