Technical Resources - Media Formulations
12491 - Advanced DMEM
ComponentsMolecular WeightConcentration (mg/L)mM
Amino Acids
Glycine75.037.50.5
L-Alanine8.9Infinity
L-Arginine hydrochloride84.0Infinity
L-Asparagine13.2Infinity
L-Aspartic acid13.3Infinity
L-Cystine 2HCl63.0Infinity
L-Glutamic Acid14.7Infinity
L-Histidine hydrochloride-H2O42.0Infinity
L-Isoleucine105.0Infinity
L-Leucine105.0Infinity
L-Lysine hydrochloride146.0Infinity
L-Methionine30.0Infinity
L-Phenylalanine66.0Infinity
L-Proline11.5Infinity
L-Serine52.5Infinity
L-Threonine95.0Infinity
L-Tryptophan16.0Infinity
L-Tyrosine disodium salt dihydrate104.0Infinity
L-Valine94.0Infinity
Vitamins
Ascorbic Acid phosphate2.5Infinity
Choline chloride4.0Infinity
D-Calcium pantothenate477.04.00.008385744
Folic Acid441.04.00.009070295
Niacinamide4.0Infinity
Pyridoxine hydrochloride4.0Infinity
Riboflavin0.4Infinity
Thiamine hydrochloride4.0Infinity
i-Inositol7.2Infinity
Inorganic Salts
Calcium Chloride (CaCl2) (anhyd.)111.0200.01.8018018
Ferric Nitrate (Fe(NO3)3"9H2O)0.1Infinity
Magnesium Sulfate (MgSO4) (anhyd.)97.67Infinity
Potassium Chloride (KCl)400.0Infinity
Sodium Bicarbonate (NaHCO3)3700.0Infinity
Sodium Chloride (NaCl)6400.0Infinity
Sodium Phosphate dibasic (Na2HPO4-H2O)125.0Infinity
Proteins
AlbuMAX® II400.0Infinity
Human Transferrin (Holo)7.5Infinity
Insulin Recombinant Full Chain10.0Infinity
Trace Elements
Ammonium Metavanadate3.0E-4Infinity
Cupric Sulfate0.00125Infinity
Manganous Chloride5.0E-5Infinity
Sodium Selenite0.005Infinity
Other Components
D-Glucose (Dextrose)180.04500.025.0
Ethanolamine1.9Infinity
Glutathione (reduced)307.01.00.0032573289
Phenol Red15.0Infinity
Sodium Pyruvate110.0Infinity
Referencia
Related Products:
12492 - Advanced MEM
Cat No: 12492013 , 12492021
12633 - Advanced RPMI 1640
Cat No: 12633012 , 12633020
12634 - Advanced D-MEM/F-12
Cat No: 12634010 , 12634028