Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Amino Acids | |||
| L-Arginine hydrochloride | 211.0 | 126.0 | 0.5971564 |
| L-Cystine 2HCl | 313.0 | 31.0 | 0.09904154 |
| L-Histidine hydrochloride-H2O | 210.0 | 42.0 | 0.2 |
| L-Isoleucine | 131.0 | 52.0 | 0.39694658 |
| L-Leucine | 131.0 | 52.0 | 0.39694658 |
| L-Lysine hydrochloride | 183.0 | 73.0 | 0.3989071 |
| L-Methionine | 149.0 | 15.0 | 0.10067114 |
| L-Phenylalanine | 165.0 | 32.0 | 0.19393939 |
| L-Threonine | 119.0 | 48.0 | 0.40336135 |
| L-Tryptophan | 204.0 | 10.0 | 0.04901961 |
| L-Tyrosine disodium salt dihydrate | 261.0 | 52.0 | 0.19923371 |
| L-Valine | 117.0 | 46.0 | 0.3931624 |
| Vitamins | |||
| Choline chloride | 140.0 | 1.0 | 0.007142857 |
| D-Calcium pantothenate | 477.0 | 1.0 | 0.002096436 |
| Folic Acid | 441.0 | 1.0 | 0.0022675737 |
| Niacinamide | 122.0 | 1.0 | 0.008196721 |
| Pyridoxal hydrochloride | 204.0 | 1.0 | 0.004901961 |
| Riboflavin | 376.0 | 0.1 | 2.6595744E-4 |
| Thiamine hydrochloride | 337.0 | 1.0 | 0.002967359 |
| i-Inositol | 180.0 | 2.0 | 0.011111111 |
| Inorganic Salts | |||
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 200.0 | 1.8018018 |
| Magnesium Sulfate (MgSO4) (anhyd.) | 120.0 | 97.67 | 0.8139166 |
| Potassium Chloride (KCl) | 75.0 | 400.0 | 5.3333335 |
| Sodium Bicarbonate (NaHCO3) | 84.0 | 2200.0 | 26.190475 |
| Sodium Chloride (NaCl) | 58.0 | 6800.0 | 117.24138 |
| Sodium Phosphate monobasic (NaH2PO4-H2O) | 138.0 | 140.0 | 1.0144928 |
| Other Components | |||
| D-Glucose (Dextrose) | 180.0 | 1000.0 | 5.5555553 |
11430 - MEM, 10x, no glutamine
Cat No:
11430030
10370 - MEM, NEAA, no glutamine
Cat No:
10370021
,
10370088
12360 - MEM, HEPES, no glutamine
Cat No:
12360038
21430 - MEM, 10x, no glutamine
Cat No:
21430020
,
21430079
32360 - MEM, HEPES, no glutamine
Cat No:
32360026
,
32360034
19993 - MEM Rega-3
Cat No:
19993013