Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Amino Acids | |||
| Glycine | 75.0 | 50.0 | 0.6666667 |
| L-Arginine hydrochloride | 211.0 | 75.0 | 0.35545024 |
| L-Aspartic acid | 133.0 | 60.0 | 0.45112783 |
| L-Cysteine | 121.0 | 61.0 | 0.5041322 |
| L-Cystine | 240.0 | 15.0 | 0.0625 |
| L-Glutamic Acid | 147.0 | 150.0 | 1.0204082 |
| L-Glutamine | 146.0 | 350.0 | 2.3972602 |
| L-Histidine | 155.0 | 128.0 | 0.82580644 |
| L-Isoleucine | 131.0 | 25.0 | 0.1908397 |
| L-Leucine | 131.0 | 50.0 | 0.3816794 |
| L-Lysine hydrochloride | 183.0 | 240.0 | 1.3114754 |
| L-Methionine | 149.0 | 50.0 | 0.33557048 |
| L-Phenylalanine | 165.0 | 50.0 | 0.3030303 |
| L-Proline | 115.0 | 50.0 | 0.4347826 |
| L-Threonine | 119.0 | 75.0 | 0.6302521 |
| L-Tryptophan | 204.0 | 40.0 | 0.19607843 |
| L-Tyrosine | 181.0 | 40.0 | 0.22099447 |
| L-Valine | 117.0 | 65.0 | 0.5555556 |
| Vitamins | |||
| Ascorbic Acid | 176.0 | 17.5 | 0.09943182 |
| Biotin | 244.0 | 0.02 | 8.196721E-5 |
| Choline chloride | 140.0 | 250.0 | 1.7857143 |
| D-Calcium pantothenate | 477.0 | 1.0 | 0.002096436 |
| Folic Acid | 441.0 | 0.5 | 0.0011337869 |
| Niacinamide | 122.0 | 1.0 | 0.008196721 |
| Pyridoxine hydrochloride | 206.0 | 1.0 | 0.004854369 |
| Riboflavin | 376.0 | 1.0 | 0.0026595744 |
| Thiamine hydrochloride | 337.0 | 10.0 | 0.029673591 |
| Vitamin B12 | 1355.0 | 0.2 | 1.4760147E-4 |
| i-Inositol | 180.0 | 1.0 | 0.0055555557 |
| Inorganic Salts | |||
| Calcium Chloride (CaCl2-2H2O) | 147.0 | 120.0 | 0.81632656 |
| Magnesium Chloride (MgCl2-6H2O) | 203.0 | 240.0 | 1.182266 |
| Magnesium Sulfate (MgSO4-7H2O) | 246.0 | 200.0 | 0.8130081 |
| Potassium Chloride (KCl) | 75.0 | 150.0 | 2.0 |
| Potassium Phosphate monobasic (KH2PO4) | 136.0 | 80.0 | 0.5882353 |
| Sodium Bicarbonate (NaHCO3) | 84.0 | 2240.0 | 26.666666 |
| Sodium Chloride (NaCl) | 58.0 | 6000.0 | 103.44827 |
| Sodium Phosphate dibasic (Na2HPO4-7H2O) | 268.0 | 566.0 | 2.1119404 |
| Other Components | |||
| D-Glucose (Dextrose) | 180.0 | 5000.0 | 27.777779 |
| Glutathione (reduced) | 307.0 | 15.0 | 0.048859935 |
| Hypoxanthine | 136.0 | 25.0 | 0.18382353 |
| Phenol Red | 376.4 | 10.0 | 0.026567481 |
1. Waymouth, C. (1968) personal communication.
31220 - Waymouth's
Cat No:
31220023
,
31220072