Technical Resources - Media Formulations
14175 - HBSS, no calcium, no magnesium, no phenol red
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Potassium Chloride (KCl)75.0400.05.3333335
Potassium Phosphate monobasic (KH2PO4)136.060.00.44117647
Sodium Bicarbonate (NaHCO3)84.0350.04.1666665
Sodium Chloride (NaCl)58.08000.0137.93103
Sodium Phosphate dibasic (Na2HPO4) anhydrous142.048.00.33802816
Other Components
D-Glucose (Dextrose)180.01000.05.5555553
Reference:
1. Hanks, J.H. and Wallace, R.E. (1949) Relation of Oxygen and Temperature in the Preservation of Tissues by Refrigeration. Proc. Soc. Exp. Biol. Med., 71:196.
Related Products:
14170 - HBSS, no calcium, no magnesium
Cat No: 14170070 , 14170088 , 14170112 , 14170120 , 14170138 , 14170146 , 14170161
14185 - HBSS, 10x, no calcium, no magnesium, no phenol red
Cat No: 14185045 , 14185052