Technical Resources - Media Formulations
21127 - Ham's F-12K (Kaighn's)
ComponentsMolecular WeightConcentration (mg/L)mM
Amino Acids
Glycine75.015.00.2
L-Alanine89.018.00.20224719
L-Arginine hydrochloride211.0422.02.0
L-Asparagine-H2O150.030.00.2
L-Aspartic acid133.026.60.2
L-Cysteine hydrochloride-H2O176.070.00.39772728
L-Glutamic Acid147.029.00.19727892
L-Glutamine146.0292.02.0
L-Histidine hydrochloride-H2O210.045.80.21809523
L-Isoleucine131.07.880.060152672
L-Leucine131.026.20.2
L-Lysine hydrochloride183.073.00.3989071
L-Methionine149.08.960.06013423
L-Phenylalanine165.09.920.060121212
L-Proline115.069.00.6
L-Serine105.021.00.2
L-Threonine119.023.00.19327731
L-Tryptophan204.04.10.020098038
L-Tyrosine disodium salt dihydrate262.013.50.051526718
L-Valine117.023.00.1965812
Vitamins
Biotin244.00.072.8688525E-4
Choline chloride140.014.00.1
D-Calcium pantothenate477.00.50.001048218
Folic Acid441.01.30.0029478457
Niacinamide122.00.0373.032787E-4
Pyridoxine hydrochloride206.00.062.9126214E-4
Riboflavin376.00.041.0638298E-4
Thiamine hydrochloride337.00.38.9020777E-4
Vitamin B121355.01.40.0010332103
i-Inositol180.018.00.1
Inorganic Salts
Calcium Chloride (CaCl2) (anhyd.)111.0102.00.9189189
Cupric sulfate (CuSO4-5H2O)250.00.0028.0E-6
Ferric sulfate (FeSO4-7H2O)278.00.80.0028776978
Magnesium Chloride (anhydrous)95.049.70.5231579
Magnesium Sulfate (MgSO4) (anhyd.)120.0192.01.6
Potassium Chloride (KCl)75.0285.03.8
Sodium Bicarbonate (NaHCO3)84.02500.029.761906
Sodium Chloride (NaCl)58.07530.0129.82759
Sodium Phosphate dibasic (Na2HPO4) anhydrous142.0115.50.8133803
Sodium Phosphate monobasic (NaH2PO4) anhydrous139.059.00.42446044
Zinc sulfate (ZnSO4-7H2O)288.00.1444.9999997E-4
Other Components
D-Glucose (Dextrose)180.01260.07.0
Hypoxanthine Na131.04.00.030534351
Lipoic Acid206.00.210.0010194174
Phenol Red376.43.00.007970245
Putrescine 2HCl161.00.320.0019875776
Sodium Pyruvate110.0220.02.0
Thymidine242.00.70.002892562
Referencia
Related Products: