Search
Halides are binary compounds formed by combining a metal with one of the five halogen elements (chlorine, bromine, fluorine, iodine, and astatine). Many of these compounds will dissolve in water, and because of this solubility, they usually occur only under special conditions. The most common halide compound is sodium chloride (NaCl), also known as rock salt.
Compared to standard wet chemical techniques, ion chromatography (IC) offers improved speed and accuracy for halide analysis. Using suppressed conductivity lowers the background conductivity caused by the eluent and effectively increases the conductivity of the analyte. Common inorganic anions such as fluoride, chloride, bromide, and sulfate can be resolved in the same injection, as well as common organic anions.
| Application Note | Sample | MDL | IC Column |
|---|---|---|---|
| Determination of Iodide in Seawater and Other Saline Matrices Using a RFIC with Suppressed Conductivity and UV Detection (AN 239) | Sea water | 15µg/L | Dionex IonPac AS20 IC Columns |
| Determination of Chloride in Acid Copper Plating Bath (AU 143) | Copper plating | 6mg/L | Dionex IonPac AS15 IC Columns |
| Quantification of Anions in Pharmaceuticals (AN 116) | Pharmaceuticals | 30–400µg/L | Dionex IonPac AS11 IC Columns |
| Ion Chromatography Assay for Chloride and Sulfate in Adenosine (AN 1075) | Pharmaceutical | 5µg/L | Dionex IonPac AS18 IC Columns |
| Determination of fluoride in sodium fluoride oral solution using carbonate/bicarbonate eluent (AN 2344) | - | - | IonPac AS22 |
| Determination of fluoride in sodium fluoride oral solution (AN 2345) | - | - | IonPac AS22 |