Search
Improve data quality, lower costs, and increase LC-MS throughput without compromising data quality or sensitivity. The Thermo Scientific Transcend systems combine proprietary Thermo Scientific TurboFlow online sample preparation technology and a unique HPLC multichannel technique to bring the productivity of up to four separate, parallel UHPLC channels to a single mass spectrometer. This reduces sample preparation time up to 95% and maximizes utilization of MS instruments.
Standard LC-MS workflows are often challenged by limited productivity due to limited throughput or extensive sample preparation steps. Transcend systems can be configured to overcome both of these limitations. As a result, your laboratory gains increased throughput by up to 400% using the multichannel Transcend LX systems. Also, automated sample clean-up is possible using the multichannel TurboFlow Transcend TLX systems.
Use a two or four channel LC system in combination with one single mass spectrometer to increase LC-MS throughput, faster return on investment, and more efficient bench space use.
Vanquish Duo for Transcend Duo LX-2 Systems Transcend Multichannel LX Systems
Use up to four channels to automate your sample cleanup with Thermo Scientific TurboFlow technology.
Use a two or four channel LC system in combination with one single mass spectrometer to increase LC-MS throughput, faster return on investment, and more efficient bench space use.
Vanquish Duo for Transcend Duo LX-2 Systems Transcend Multichannel LX Systems
Use up to four channels to automate your sample cleanup with Thermo Scientific TurboFlow technology.