Search
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
| 2 | 633 | 780/60 | 756 | 785 | (in buffer) 1 | flow cytometry |
Invitrogen APC-eFluor 780 dye is a near-infrared fluorescent dye designed to be a high-performance tandem fluorophore for multicolor flow cytometry. It is designed for use with flow cytometers equipped with a red laser and have an emission peak of 780 nm. With similar spectral characteristics to APC-Alexa Fluor 750, APC-eFluor 780 requires no changes to your instrument configuration for use, allowing you to easily add APC-eFluor 780 to your multicolor panels.
We offer APC-eFluor 780 dye conjugated to primary antibodies for use in flow cytometry.
Comparison of APC-eFluor 780 dye to APC-H7 dye using flow cytometry. Mouse splenocytes were stained with CD8a Monoclonal Antibody (53-6.7), APC-eFluor 780 (blue line) or CD8a monoclonal antibody conjugated to APC-H7 (red line).
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
A handy reference poster featuring the broad range of our available dyes and labeling reagents.
Protocols that fit your needs in imaging ranging from sample and assay preparation to staining, labeling, and data analysis strategies.
A tool for selecting the optimal fluorescent dyes for your Invitrogen EVOS cell imaging systems.
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.