Search
The uncompromised quality of Thermo Scientific B·R·A·H·M·S KRYPTOR Analyzers supports relevant clinical decisions for your laboratory.
B·R·A·H·M·S KRYPTOR Analyzers support safe patient management with:
| BꞏRꞏAꞏHꞏMꞏS KRYPTOR Analyzers | ![]() |
![]() |
| Specifications | B·R·A·H·M·S KRYPTOR GOLD | |
| Dimensions (W x H x D) | 93 x 73 (121 with tower light or with open hood) x 72 cm | 74 x 63 x 75 cm |
| Weight | 118 kg | 54 kg |
| Electrical consumption | 465 W | 700 W |
Throughput (per hour) |
Up to 115 tests | Up to 60 tests |
| Sample capacity | Up to 80 samples | Up to 64 samples |
| Reagent capacity | Up to 16 kits | Up to 8 kits |
Thermo Fisher Scientific products are distributed globally and their uses, applications, indications, claims and availability of products in each country depend on local regulatory marketing authorization status, please consult the Instructions For Use (IFU) available in your country.
© 2024 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified. B·R·A·H·M·S is a registered trademark of B·R·A·H·M·S GmbH.
KRYPTOR and TRACE are trademarks of Cisbio Bioassays, licensed for use by B·R·A·H·M·S GmbH, a part of Thermo Fisher Scientific. Nobel Prize is a registered trademark of the Nobel Foundation.